Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 43669971-43670240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLAUR
Entrez Gene ID
5329
Gene Name
plasminogen activator, urokinase receptor
Gene Aliases
CD87, U-PAR, UPAR, URKR
UniGene
Hs.466871
Gene Chromosome Location
Chr.19: 43646095-43670346 on Build GRCh38
Transcripts
NM_001005376.2 NM_001005377.2 NM_001301037.1 NM_002659.3 XM_005258990.4 XM_011527027.2 XM_011527028.2 XM_011527029.1 XM_011527030.2 XM_011527031.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4078985 c.22C>T p.P8S Chr.19: 43670099-43670099 on Build GRCh38
997688 c.26T>C p.L9P Chr.19: 43670095-43670095 on Build GRCh38
4754058 c.29T>C p.L10P Chr.19: 43670092-43670092 on Build GRCh38
4754057 c.29T>C p.L10P Chr.19: 43670092-43670092 on Build GRCh38
4593186 c.8A>C p.H3P Chr.19: 43670113-43670113 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774527050 Chr.19: 43670083-43670088 on Build GRCh38
rs377296256 Chr.19: 43670156-43670156 on Build GRCh38
rs1013238770 Chr.19: 43670211-43670211 on Build GRCh38
rs755139842 Chr.19: 43670089-43670089 on Build GRCh38
rs369947389 Chr.19: 43670029-43670029 on Build GRCh38
rs745774333 Chr.19: 43670113-43670113 on Build GRCh38
rs1032820404 Chr.19: 43670004-43670004 on Build GRCh38
rs147665588 Chr.19: 43670207-43670207 on Build GRCh38
rs36065389 Chr.19: 43670216-43670217 on Build GRCh38
rs766889369 Chr.19: 43670161-43670161 on Build GRCh38
rs201156019 Chr.19: 43670106-43670106 on Build GRCh38
rs761181656 Chr.19: 43670168-43670170 on Build GRCh38
rs200437703 Chr.19: 43670060-43670060 on Build GRCh38
rs773722571 Chr.19: 43670167-43670174 on Build GRCh38
rs115313724 Chr.19: 43669997-43669997 on Build GRCh38
rs761914709 Chr.19: 43670090-43670098 on Build GRCh38
rs779258618 Chr.19: 43670050-43670050 on Build GRCh38
rs767537419 Chr.19: 43670128-43670145 on Build GRCh38
rs976215233 Chr.19: 43670046-43670046 on Build GRCh38
rs751627526 Chr.19: 43670168-43670168 on Build GRCh38
rs750990516 Chr.19: 43670054-43670054 on Build GRCh38
rs947179518 Chr.19: 43670108-43670108 on Build GRCh38
rs952193763 Chr.19: 43670006-43670006 on Build GRCh38
rs771905230 Chr.19: 43670120-43670120 on Build GRCh38
rs373838084 Chr.19: 43670030-43670030 on Build GRCh38
rs780069481 Chr.19: 43670076-43670076 on Build GRCh38
rs914209076 Chr.19: 43670099-43670099 on Build GRCh38
rs760336050 Chr.19: 43670028-43670028 on Build GRCh38
rs552297751 Chr.19: 43670026-43670026 on Build GRCh38
rs923520441 Chr.19: 43670049-43670049 on Build GRCh38
rs781453492 Chr.19: 43670091-43670091 on Build GRCh38
rs924480029 Chr.19: 43670150-43670150 on Build GRCh38
rs765588443 Chr.19: 43670147-43670147 on Build GRCh38
rs757706098 Chr.19: 43670039-43670039 on Build GRCh38
rs763833064 Chr.19: 43670128-43670128 on Build GRCh38
rs17559065 Chr.19: 43670219-43670219 on Build GRCh38
rs575942082 Chr.19: 43670139-43670139 on Build GRCh38
rs62639324 Chr.19: 43670097-43670097 on Build GRCh38
rs920741471 Chr.19: 43670218-43670218 on Build GRCh38
rs750785227 Chr.19: 43670153-43670153 on Build GRCh38
rs554683003 Chr.19: 43670079-43670079 on Build GRCh38
rs1000416994 Chr.19: 43669998-43669998 on Build GRCh38
rs768969786 Chr.19: 43670083-43670091 on Build GRCh38
rs117499826 Chr.19: 43670124-43670124 on Build GRCh38
rs775082136 Chr.19: 43670123-43670123 on Build GRCh38
rs867018678 Chr.19: 43670098-43670098 on Build GRCh38
rs776464186 Chr.19: 43670138-43670138 on Build GRCh38

View Full Product Details