Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 51689228-51689485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCG3
Entrez Gene ID
29106
Gene Name
secretogranin III
Gene Aliases
SGIII
UniGene
Hs.232618
Gene Chromosome Location
Chr.15: 51681353-51721026 on Build GRCh38
Transcripts
NM_001165257.1 NM_013243.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5538561 c.653G>A p.W218* Chr.15: 51689331-51689331 on Build GRCh38
5383486 c.550A>T p.S184C Chr.15: 51689228-51689228 on Build GRCh38
4555885 c.671G>A p.G224E Chr.15: 51689349-51689349 on Build GRCh38
4755203 c.555A>T p.Q185H Chr.15: 51689233-51689233 on Build GRCh38
4556422 c.690G>A p.V230V Chr.15: 51689368-51689368 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs955231129 Chr.15: 51689314-51689314 on Build GRCh38
rs763648244 Chr.15: 51689399-51689408 on Build GRCh38
rs761803593 Chr.15: 51689397-51689400 on Build GRCh38
rs867041137 Chr.15: 51689446-51689446 on Build GRCh38
rs555096648 Chr.15: 51689317-51689317 on Build GRCh38
rs189719461 Chr.15: 51689308-51689308 on Build GRCh38
rs774538301 Chr.15: 51689396-51689397 on Build GRCh38
rs200742282 Chr.15: 51689402-51689402 on Build GRCh38
rs749520347 Chr.15: 51689313-51689313 on Build GRCh38
rs749266652 Chr.15: 51689406-51689406 on Build GRCh38
rs779900108 Chr.15: 51689294-51689294 on Build GRCh38
rs760171204 Chr.15: 51689399-51689406 on Build GRCh38
rs182182990 Chr.15: 51689338-51689338 on Build GRCh38
rs774789007 Chr.15: 51689327-51689327 on Build GRCh38
rs796362728 Chr.15: 51689399-51689400 on Build GRCh38
rs763631511 Chr.15: 51689354-51689354 on Build GRCh38
rs750223376 Chr.15: 51689391-51689391 on Build GRCh38
rs62016039 Chr.15: 51689365-51689365 on Build GRCh38
rs772399547 Chr.15: 51689399-51689404 on Build GRCh38
rs201709954 Chr.15: 51689399-51689402 on Build GRCh38
rs1038691618 Chr.15: 51689401-51689404 on Build GRCh38
rs776215434 Chr.15: 51689369-51689369 on Build GRCh38
rs776237354 Chr.15: 51689397-51689398 on Build GRCh38
rs756163980 Chr.15: 51689296-51689296 on Build GRCh38
rs537976380 Chr.15: 51689353-51689353 on Build GRCh38
rs764741001 Chr.15: 51689388-51689388 on Build GRCh38
rs79392293 Chr.15: 51689441-51689441 on Build GRCh38
rs929610961 Chr.15: 51689381-51689381 on Build GRCh38
rs3841591 Chr.15: 51689425-51689432 on Build GRCh38
rs557583431 Chr.15: 51689373-51689373 on Build GRCh38
rs1055957720 Chr.15: 51689411-51689411 on Build GRCh38
rs762618970 Chr.15: 51689398-51689399 on Build GRCh38
rs142654979 Chr.15: 51689293-51689293 on Build GRCh38
rs3223422 Chr.15: 51689372-51689373 on Build GRCh38
rs766353264 Chr.15: 51689397-51689397 on Build GRCh38
rs71426081 Chr.15: 51689401-51689402 on Build GRCh38
rs745575095 Chr.15: 51689399-51689400 on Build GRCh38
rs796858791 Chr.15: 51689397-51689398 on Build GRCh38
rs751467315 Chr.15: 51689399-51689399 on Build GRCh38
rs758346562 Chr.15: 51689284-51689284 on Build GRCh38
rs748249995 Chr.15: 51689295-51689295 on Build GRCh38
rs756250388 Chr.15: 51689410-51689411 on Build GRCh38
rs918293448 Chr.15: 51689401-51689412 on Build GRCh38
rs756214486 Chr.15: 51689401-51689401 on Build GRCh38
rs758150258 Chr.15: 51689393-51689393 on Build GRCh38
rs750139019 Chr.15: 51689400-51689401 on Build GRCh38
rs112308294 Chr.15: 51689322-51689464 on Build GRCh38
rs757503292 Chr.15: 51689407-51689407 on Build GRCh38
rs68133708 Chr.15: 51689400-51689403 on Build GRCh38
rs985282299 Chr.15: 51689258-51689258 on Build GRCh38
rs776096473 Chr.15: 51689344-51689344 on Build GRCh38
rs1456301 Chr.15: 51689400-51689400 on Build GRCh38

View Full Product Details