Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 25251908-25252139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTMR6
Entrez Gene ID
9107
Gene Name
myotubularin related protein 6
Gene Aliases
-
UniGene
Hs.643702
Gene Chromosome Location
Chr.13: 25246201-25287566 on Build GRCh38
Transcripts
NM_004685.3 XM_011535306.1 XM_011535307.1 XM_017020845.1 XM_017020846.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4916616 c.1413T>A p.P471P Chr.13: 25251918-25251918 on Build GRCh38
946292 c.1398G>T p.K466N Chr.13: 25251933-25251933 on Build GRCh38
6138882 c.1397A>C p.K466T Chr.13: 25251934-25251934 on Build GRCh38
6410718 c.1388A>G p.E463G Chr.13: 25251943-25251943 on Build GRCh38
5970946 c.1386G>A p.L462L Chr.13: 25251945-25251945 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747363693 Chr.13: 25252025-25252025 on Build GRCh38
rs778454169 Chr.13: 25252012-25252012 on Build GRCh38
rs774413920 Chr.13: 25251980-25251980 on Build GRCh38
rs1022078859 Chr.13: 25252079-25252079 on Build GRCh38
rs765453389 Chr.13: 25251972-25251972 on Build GRCh38
rs372446234 Chr.13: 25252020-25252020 on Build GRCh38
rs749722751 Chr.13: 25251946-25251952 on Build GRCh38
rs76317728 Chr.13: 25251945-25251945 on Build GRCh38
rs967886109 Chr.13: 25252103-25252103 on Build GRCh38
rs753572087 Chr.13: 25252004-25252004 on Build GRCh38
rs1018241220 Chr.13: 25252066-25252066 on Build GRCh38
rs773868314 Chr.13: 25252075-25252075 on Build GRCh38
rs376864350 Chr.13: 25252085-25252085 on Build GRCh38
rs771366436 Chr.13: 25252029-25252029 on Build GRCh38
rs748650975 Chr.13: 25252014-25252014 on Build GRCh38
rs776731789 Chr.13: 25251953-25251953 on Build GRCh38
rs771448432 Chr.13: 25252003-25252004 on Build GRCh38
rs140082792 Chr.13: 25251966-25251966 on Build GRCh38
rs778061798 Chr.13: 25252019-25252019 on Build GRCh38
rs770216169 Chr.13: 25251976-25251976 on Build GRCh38
rs750202009 Chr.13: 25251995-25251995 on Build GRCh38
rs758305289 Chr.13: 25252046-25252046 on Build GRCh38
rs117113767 Chr.13: 25252076-25252076 on Build GRCh38
rs138289796 Chr.13: 25251950-25251950 on Build GRCh38
rs767351906 Chr.13: 25251991-25251991 on Build GRCh38
rs765983201 Chr.13: 25252000-25252000 on Build GRCh38
rs761906117 Chr.13: 25251981-25251981 on Build GRCh38
rs776691722 Chr.13: 25252036-25252036 on Build GRCh38
rs758912334 Chr.13: 25252016-25252016 on Build GRCh38
rs754462109 Chr.13: 25252005-25252005 on Build GRCh38
rs771079522 Chr.13: 25251944-25251944 on Build GRCh38
rs755746839 Chr.13: 25251996-25251996 on Build GRCh38

View Full Product Details