Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2759229-2759490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SRRM2
Entrez Gene ID
23524
Gene Name
serine/arginine repetitive matrix 2
Gene Aliases
300-KD, CWF21, Cwc21, HSPC075, SRL300, SRm300
UniGene
Hs.433343
Gene Chromosome Location
Chr.16: 2752329-2771412 on Build GRCh38
Transcripts
NM_016333.3 XM_005255226.1 XM_006720874.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1227583 c.739C>T p.R247W Chr.16: 2759401-2759401 on Build GRCh38
4059713 c.733C>T p.R245* Chr.16: 2759395-2759395 on Build GRCh38
3690886 c.734G>A p.R245Q Chr.16: 2759396-2759396 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs946200782 Chr.16: 2759433-2759433 on Build GRCh38
rs772221460 Chr.16: 2759360-2759360 on Build GRCh38
rs544572991 Chr.16: 2759341-2759341 on Build GRCh38
rs781006645 Chr.16: 2759325-2759325 on Build GRCh38
rs762505544 Chr.16: 2759307-2759307 on Build GRCh38
rs747331898 Chr.16: 2759374-2759374 on Build GRCh38
rs777924298 Chr.16: 2759345-2759345 on Build GRCh38
rs756176894 Chr.16: 2759423-2759423 on Build GRCh38
rs751224645 Chr.16: 2759317-2759317 on Build GRCh38
rs113294042 Chr.16: 2759261-2759261 on Build GRCh38
rs769028384 Chr.16: 2759375-2759375 on Build GRCh38
rs747274700 Chr.16: 2759449-2759449 on Build GRCh38
rs765849114 Chr.16: 2759308-2759308 on Build GRCh38
rs1015496573 Chr.16: 2759455-2759455 on Build GRCh38
rs1028036014 Chr.16: 2759432-2759433 on Build GRCh38
rs781209469 Chr.16: 2759307-2759308 on Build GRCh38
rs368595576 Chr.16: 2759294-2759294 on Build GRCh38
rs748093655 Chr.16: 2759409-2759419 on Build GRCh38
rs190699786 Chr.16: 2759444-2759444 on Build GRCh38
rs114827582 Chr.16: 2759297-2759297 on Build GRCh38
rs754739314 Chr.16: 2759324-2759324 on Build GRCh38
rs749231720 Chr.16: 2759340-2759340 on Build GRCh38
rs578021877 Chr.16: 2759443-2759443 on Build GRCh38
rs1009264179 Chr.16: 2759367-2759367 on Build GRCh38
rs1055492882 Chr.16: 2759257-2759257 on Build GRCh38
rs777600039 Chr.16: 2759430-2759431 on Build GRCh38
rs955493620 Chr.16: 2759347-2759347 on Build GRCh38
rs752421519 Chr.16: 2759420-2759420 on Build GRCh38
rs752474487 Chr.16: 2759333-2759333 on Build GRCh38
rs763833569 Chr.16: 2759430-2759430 on Build GRCh38
rs111721270 Chr.16: 2759405-2759405 on Build GRCh38
rs764515417 Chr.16: 2759302-2759302 on Build GRCh38
rs754330004 Chr.16: 2759306-2759306 on Build GRCh38
rs76786072 Chr.16: 2759445-2759445 on Build GRCh38
rs778926980 Chr.16: 2759355-2759355 on Build GRCh38
rs775690276 Chr.16: 2759361-2759361 on Build GRCh38
rs778919400 Chr.16: 2759435-2759435 on Build GRCh38
rs938326612 Chr.16: 2759254-2759254 on Build GRCh38
rs368275451 Chr.16: 2759450-2759450 on Build GRCh38
rs777167753 Chr.16: 2759387-2759387 on Build GRCh38
rs755953001 Chr.16: 2759424-2759424 on Build GRCh38
rs938805059 Chr.16: 2759336-2759336 on Build GRCh38
rs908437160 Chr.16: 2759272-2759272 on Build GRCh38
rs747144306 Chr.16: 2759432-2759432 on Build GRCh38
rs777718531 Chr.16: 2759339-2759339 on Build GRCh38
rs762296235 Chr.16: 2759392-2759392 on Build GRCh38
rs756006206 Chr.16: 2759334-2759334 on Build GRCh38
rs754557612 Chr.16: 2759302-2759307 on Build GRCh38
rs990813132 Chr.16: 2759417-2759417 on Build GRCh38
rs773680637 Chr.16: 2759397-2759397 on Build GRCh38
rs138293757 Chr.16: 2759395-2759395 on Build GRCh38
rs374586831 Chr.16: 2759258-2759258 on Build GRCh38
rs202002640 Chr.16: 2759401-2759401 on Build GRCh38
rs767239423 Chr.16: 2759411-2759411 on Build GRCh38
rs536153993 Chr.16: 2759275-2759275 on Build GRCh38
rs879705289 Chr.16: 2759464-2759464 on Build GRCh38
rs372623157 Chr.16: 2759358-2759358 on Build GRCh38
rs1018257369 Chr.16: 2759396-2759396 on Build GRCh38

View Full Product Details