Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 179375121-179375348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFN1
Entrez Gene ID
55669
Gene Name
mitofusin 1
Gene Aliases
hfzo1, hfzo2
UniGene
Hs.478383
Gene Chromosome Location
Chr.3: 179347692-179394944 on Build GRCh38
Transcripts
NM_033540.2 XM_005247596.3 XM_011512963.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
276064 c.1011C>T p.F337F Chr.3: 179375255-179375255 on Build GRCh38
240660 c.1070C>T p.S357L Chr.3: 179375314-179375314 on Build GRCh38
1041544 c.1086T>C p.A362A Chr.3: 179375330-179375330 on Build GRCh38
5863969 c.995C>T p.A332V Chr.3: 179375239-179375239 on Build GRCh38
4115440 c.1012G>A p.E338K Chr.3: 179375256-179375256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755879355 Chr.3: 179375230-179375230 on Build GRCh38
rs751976234 Chr.3: 179375256-179375256 on Build GRCh38
rs756260332 Chr.3: 179375276-179375276 on Build GRCh38
rs372512172 Chr.3: 179375253-179375253 on Build GRCh38
rs978869700 Chr.3: 179375309-179375309 on Build GRCh38
rs753978980 Chr.3: 179375292-179375292 on Build GRCh38
rs750993218 Chr.3: 179375178-179375178 on Build GRCh38
rs781514055 Chr.3: 179375202-179375202 on Build GRCh38
rs777965394 Chr.3: 179375286-179375286 on Build GRCh38
rs779955116 Chr.3: 179375216-179375216 on Build GRCh38
rs777770805 Chr.3: 179375210-179375210 on Build GRCh38
rs200698063 Chr.3: 179375207-179375207 on Build GRCh38
rs376997527 Chr.3: 179375268-179375268 on Build GRCh38
rs968763949 Chr.3: 179375304-179375305 on Build GRCh38
rs752015975 Chr.3: 179375194-179375194 on Build GRCh38
rs745923221 Chr.3: 179375247-179375248 on Build GRCh38
rs772783862 Chr.3: 179375250-179375250 on Build GRCh38
rs200775244 Chr.3: 179375227-179375227 on Build GRCh38
rs370622566 Chr.3: 179375203-179375203 on Build GRCh38
rs371331572 Chr.3: 179375204-179375205 on Build GRCh38
rs774309739 Chr.3: 179375215-179375215 on Build GRCh38
rs780081711 Chr.3: 179375314-179375314 on Build GRCh38
rs766857292 Chr.3: 179375189-179375189 on Build GRCh38
rs769445078 Chr.3: 179375231-179375231 on Build GRCh38
rs543443619 Chr.3: 179375255-179375255 on Build GRCh38
rs149864152 Chr.3: 179375223-179375223 on Build GRCh38
rs764804098 Chr.3: 179375177-179375177 on Build GRCh38
rs528951205 Chr.3: 179375319-179375319 on Build GRCh38
rs1030476572 Chr.3: 179375212-179375212 on Build GRCh38
rs9865666 Chr.3: 179375201-179375201 on Build GRCh38
rs200339629 Chr.3: 179375265-179375265 on Build GRCh38
rs757306285 Chr.3: 179375310-179375310 on Build GRCh38
rs746871851 Chr.3: 179375217-179375217 on Build GRCh38
rs745808096 Chr.3: 179375213-179375213 on Build GRCh38

View Full Product Details