Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49631757-49632030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPF40B
Entrez Gene ID
25766
Gene Name
pre-mRNA processing factor 40 homolog B
Gene Aliases
HYPC
UniGene
Hs.706827
Gene Chromosome Location
Chr.12: 49622717-49644669 on Build GRCh38
Transcripts
NM_001031698.2 NM_012272.2 XM_006719324.3 XM_006719325.3 XM_011538135.2 XM_011538136.2 XM_011538137.2 XM_011538138.2 XM_011538139.2 XM_011538140.2 XM_011538141.1 XM_011538143.2 XM_011538144.2 XM_017019135.1 XM_017019136.1 XM_017019137.1 XM_017019138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4775042 c.182C>T p.P61L Chr.12: 49631879-49631879 on Build GRCh38
3812188 c.163-1G>C p.? Chr.12: 49631859-49631859 on Build GRCh38
6278481 c.209C>T p.A70V Chr.12: 49631906-49631906 on Build GRCh38
3461697 c.221C>T p.T74I Chr.12: 49631918-49631918 on Build GRCh38
2008093 c.222C>T p.T74T Chr.12: 49631919-49631919 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763707488 Chr.12: 49631911-49631911 on Build GRCh38
rs375767554 Chr.12: 49631801-49631801 on Build GRCh38
rs767628408 Chr.12: 49631906-49631906 on Build GRCh38
rs749433984 Chr.12: 49631835-49631835 on Build GRCh38
rs927008992 Chr.12: 49631922-49631922 on Build GRCh38
rs781088130 Chr.12: 49631832-49631832 on Build GRCh38
rs751532960 Chr.12: 49631827-49631827 on Build GRCh38
rs946206565 Chr.12: 49631998-49631998 on Build GRCh38
rs758227369 Chr.12: 49631821-49631821 on Build GRCh38
rs146982042 Chr.12: 49631904-49631904 on Build GRCh38
rs771356358 Chr.12: 49631975-49631975 on Build GRCh38
rs369232303 Chr.12: 49631880-49631880 on Build GRCh38
rs750614540 Chr.12: 49631971-49631971 on Build GRCh38
rs868573920 Chr.12: 49631932-49631932 on Build GRCh38
rs370814816 Chr.12: 49631972-49631972 on Build GRCh38
rs763751338 Chr.12: 49631823-49631823 on Build GRCh38
rs757228427 Chr.12: 49631831-49631831 on Build GRCh38
rs778814805 Chr.12: 49631955-49631955 on Build GRCh38
rs750265621 Chr.12: 49631924-49631924 on Build GRCh38
rs773749747 Chr.12: 49631874-49631874 on Build GRCh38
rs761504488 Chr.12: 49631979-49631979 on Build GRCh38
rs141374337 Chr.12: 49631884-49631884 on Build GRCh38
rs781768479 Chr.12: 49631977-49631977 on Build GRCh38
rs765008162 Chr.12: 49631814-49631814 on Build GRCh38
rs890261265 Chr.12: 49631790-49631790 on Build GRCh38
rs369038774 Chr.12: 49631843-49631843 on Build GRCh38
rs201337152 Chr.12: 49631856-49631856 on Build GRCh38
rs777273691 Chr.12: 49631881-49631881 on Build GRCh38
rs752788631 Chr.12: 49631958-49631958 on Build GRCh38
rs149054908 Chr.12: 49631920-49631920 on Build GRCh38
rs377275190 Chr.12: 49631961-49631961 on Build GRCh38
rs374803402 Chr.12: 49631861-49631861 on Build GRCh38
rs752225552 Chr.12: 49631818-49631818 on Build GRCh38
rs914801213 Chr.12: 49631983-49631983 on Build GRCh38
rs762476011 Chr.12: 49631910-49631910 on Build GRCh38
rs372962598 Chr.12: 49631907-49631907 on Build GRCh38
rs147595412 Chr.12: 49631917-49631917 on Build GRCh38
rs748266938 Chr.12: 49631857-49631857 on Build GRCh38
rs974659128 Chr.12: 49631799-49631799 on Build GRCh38
rs747355839 Chr.12: 49631877-49631877 on Build GRCh38
rs755104847 Chr.12: 49631925-49631925 on Build GRCh38
rs757036317 Chr.12: 49631919-49631919 on Build GRCh38

View Full Product Details