Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1510217-1510456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTSL5
Entrez Gene ID
339366
Gene Name
ADAMTS like 5
Gene Aliases
THSD6
UniGene
Hs.371674
Gene Chromosome Location
Chr.19: 1505018-1513189 on Build GRCh38
Transcripts
NM_213604.2 XM_005259549.3 XM_011527961.1 XM_011527963.1 XM_017026744.1 XM_017026745.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6900228 c.262C>T p.P88S Chr.19: 1510249-1510249 on Build GRCh38
4942610 c.236G>A p.R79H Chr.19: 1510384-1510384 on Build GRCh38
460044 c.277T>C p.F93L Chr.19: 1510234-1510234 on Build GRCh38
5854833 c.205G>A p.E69K Chr.19: 1510415-1510415 on Build GRCh38
4074905 c.291G>A p.Q97Q Chr.19: 1510220-1510220 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199888356 Chr.19: 1510277-1510277 on Build GRCh38
rs957795150 Chr.19: 1510240-1510240 on Build GRCh38
rs768863354 Chr.19: 1510299-1510299 on Build GRCh38
rs200659727 Chr.19: 1510258-1510258 on Build GRCh38
rs558828966 Chr.19: 1510296-1510296 on Build GRCh38
rs919842775 Chr.19: 1510380-1510380 on Build GRCh38
rs745850367 Chr.19: 1510284-1510284 on Build GRCh38
rs149832091 Chr.19: 1510411-1510411 on Build GRCh38
rs781440777 Chr.19: 1510407-1510407 on Build GRCh38
rs753613108 Chr.19: 1510275-1510275 on Build GRCh38
rs202076670 Chr.19: 1510369-1510369 on Build GRCh38
rs1008987267 Chr.19: 1510352-1510352 on Build GRCh38
rs56891350 Chr.19: 1510334-1510334 on Build GRCh38
rs367836169 Chr.19: 1510337-1510337 on Build GRCh38
rs773636437 Chr.19: 1510413-1510413 on Build GRCh38
rs780346712 Chr.19: 1510296-1510296 on Build GRCh38
rs967062522 Chr.19: 1510270-1510270 on Build GRCh38
rs937294926 Chr.19: 1510346-1510346 on Build GRCh38
rs370862596 Chr.19: 1510318-1510318 on Build GRCh38
rs771371395 Chr.19: 1510424-1510424 on Build GRCh38
rs770187913 Chr.19: 1510326-1510326 on Build GRCh38
rs376576492 Chr.19: 1510268-1510268 on Build GRCh38
rs748371361 Chr.19: 1510410-1510410 on Build GRCh38
rs752185651 Chr.19: 1510357-1510357 on Build GRCh38
rs778909579 Chr.19: 1510374-1510374 on Build GRCh38
rs776736911 Chr.19: 1510300-1510300 on Build GRCh38
rs755040266 Chr.19: 1510402-1510402 on Build GRCh38
rs751921530 Chr.19: 1510247-1510248 on Build GRCh38
rs755864822 Chr.19: 1510262-1510262 on Build GRCh38
rs778962754 Chr.19: 1510278-1510278 on Build GRCh38
rs368654852 Chr.19: 1510391-1510391 on Build GRCh38
rs527986426 Chr.19: 1510298-1510298 on Build GRCh38
rs757052752 Chr.19: 1510373-1510373 on Build GRCh38
rs777297263 Chr.19: 1510269-1510269 on Build GRCh38
rs767013943 Chr.19: 1510347-1510347 on Build GRCh38
rs767067291 Chr.19: 1510252-1510252 on Build GRCh38
rs763511871 Chr.19: 1510339-1510339 on Build GRCh38
rs750379187 Chr.19: 1510379-1510379 on Build GRCh38
rs898984780 Chr.19: 1510266-1510266 on Build GRCh38
rs552944361 Chr.19: 1510370-1510370 on Build GRCh38
rs981082260 Chr.19: 1510272-1510272 on Build GRCh38
rs758342918 Chr.19: 1510384-1510384 on Build GRCh38
rs772264053 Chr.19: 1510289-1510289 on Build GRCh38
rs371584830 Chr.19: 1510362-1510362 on Build GRCh38
rs780150402 Chr.19: 1510385-1510385 on Build GRCh38
rs749660942 Chr.19: 1510423-1510423 on Build GRCh38

View Full Product Details