Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 128008923-128009152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD18
Entrez Gene ID
80167
Gene Name
abhydrolase domain containing 18
Gene Aliases
C4orf29
UniGene
Hs.143995
Gene Chromosome Location
Chr.4: 127965256-128039711 on Build GRCh38
Transcripts
NM_001319305.1 NM_001319306.1 NM_001319307.1 XM_005263238.4 XM_006714318.3 XM_006714320.3 XM_006714322.3 XM_011532272.2 XM_011532273.2 XM_011532274.2 XM_011532276.1 XM_011532278.2 XM_017008631.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1426980 c.373C>T p.R125* Chr.4: 128009122-128009122 on Build GRCh38
4437598 c.320G>C p.R107T Chr.4: 128008961-128008961 on Build GRCh38
6434307 c.379C>A p.L127I Chr.4: 128009128-128009128 on Build GRCh38
4158791 c.283C>A p.Q95K Chr.4: 128008924-128008924 on Build GRCh38
6616149 c.334C>A p.H112N Chr.4: 128008975-128008975 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776345754 Chr.4: 128009084-128009084 on Build GRCh38
rs553379780 Chr.4: 128009038-128009038 on Build GRCh38
rs763242274 Chr.4: 128009058-128009058 on Build GRCh38
rs747288381 Chr.4: 128009097-128009097 on Build GRCh38
rs762453802 Chr.4: 128009046-128009046 on Build GRCh38
rs574956758 Chr.4: 128009119-128009119 on Build GRCh38
rs377192233 Chr.4: 128009071-128009071 on Build GRCh38
rs374739409 Chr.4: 128009087-128009087 on Build GRCh38
rs769642474 Chr.4: 128009120-128009120 on Build GRCh38
rs567962998 Chr.4: 128008991-128008991 on Build GRCh38
rs368923834 Chr.4: 128009016-128009016 on Build GRCh38
rs373107703 Chr.4: 128009076-128009076 on Build GRCh38
rs771840263 Chr.4: 128009021-128009021 on Build GRCh38
rs778833440 Chr.4: 128008999-128008999 on Build GRCh38
rs375069964 Chr.4: 128008974-128008974 on Build GRCh38
rs773100732 Chr.4: 128009122-128009122 on Build GRCh38
rs981870436 Chr.4: 128009056-128009056 on Build GRCh38
rs376404847 Chr.4: 128009030-128009030 on Build GRCh38
rs766700363 Chr.4: 128009069-128009069 on Build GRCh38
rs749130165 Chr.4: 128009088-128009088 on Build GRCh38
rs747951881 Chr.4: 128009089-128009089 on Build GRCh38
rs116482971 Chr.4: 128008956-128008956 on Build GRCh38
rs373527023 Chr.4: 128009026-128009026 on Build GRCh38
rs966342717 Chr.4: 128009055-128009055 on Build GRCh38
rs763633753 Chr.4: 128009077-128009077 on Build GRCh38
rs144765403 Chr.4: 128009085-128009085 on Build GRCh38
rs538231850 Chr.4: 128009081-128009081 on Build GRCh38
rs770823501 Chr.4: 128008992-128008992 on Build GRCh38
rs952889212 Chr.4: 128008987-128008987 on Build GRCh38
rs755626067 Chr.4: 128008968-128008968 on Build GRCh38
rs753341111 Chr.4: 128009078-128009078 on Build GRCh38
rs373062649 Chr.4: 128009049-128009049 on Build GRCh38
rs867186579 Chr.4: 128008958-128008958 on Build GRCh38
rs768883079 Chr.4: 128009044-128009044 on Build GRCh38
rs776833559 Chr.4: 128009045-128009045 on Build GRCh38
rs188809510 Chr.4: 128009047-128009047 on Build GRCh38

View Full Product Details