Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127732475-127732732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOR2A
Entrez Gene ID
27433
Gene Name
torsin family 2 member A
Gene Aliases
TORP1
UniGene
Hs.444106
Gene Chromosome Location
Chr.9: 127731524-127735387 on Build GRCh38
Transcripts
NM_001085347.2 NM_001134430.2 NM_001134431.2 NM_001252018.1 NM_001252021.1 NM_001252023.1 NM_130459.3 XM_011518554.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4997012 c.656G>T p.R219L Chr.9: 127732629-127732629 on Build GRCh38
1230092 c.599C>T p.T200M Chr.9: 127732686-127732686 on Build GRCh38
7422991 c.651G>T p.R217R Chr.9: 127732634-127732634 on Build GRCh38
4735362 c.647G>A p.R216Q Chr.9: 127732638-127732638 on Build GRCh38
6248004 c.715C>A p.P239T Chr.9: 127732570-127732570 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536262 Chr.9: 127732512-127732512 on Build GRCh38
rs756975653 Chr.9: 127732537-127732537 on Build GRCh38
rs754863726 Chr.9: 127732684-127732684 on Build GRCh38
rs748293678 Chr.9: 127732576-127732576 on Build GRCh38
rs987353655 Chr.9: 127732626-127732626 on Build GRCh38
rs780944838 Chr.9: 127732541-127732541 on Build GRCh38
rs543471487 Chr.9: 127732584-127732584 on Build GRCh38
rs200609209 Chr.9: 127732563-127732563 on Build GRCh38
rs750090106 Chr.9: 127732606-127732606 on Build GRCh38
rs376325991 Chr.9: 127732667-127732667 on Build GRCh38
rs754405976 Chr.9: 127732529-127732529 on Build GRCh38
rs377208966 Chr.9: 127732700-127732700 on Build GRCh38
rs749269975 Chr.9: 127732565-127732565 on Build GRCh38
rs779854900 Chr.9: 127732627-127732627 on Build GRCh38
rs144539918 Chr.9: 127732649-127732649 on Build GRCh38
rs774385889 Chr.9: 127732517-127732517 on Build GRCh38
rs768577591 Chr.9: 127732568-127732568 on Build GRCh38
rs548368375 Chr.9: 127732650-127732650 on Build GRCh38
rs940676114 Chr.9: 127732660-127732660 on Build GRCh38
rs371270860 Chr.9: 127732629-127732629 on Build GRCh38
rs996680614 Chr.9: 127732592-127732592 on Build GRCh38
rs563260865 Chr.9: 127732586-127732586 on Build GRCh38
rs751130884 Chr.9: 127732598-127732598 on Build GRCh38
rs367989583 Chr.9: 127732644-127732644 on Build GRCh38
rs761246198 Chr.9: 127732525-127732525 on Build GRCh38
rs199671418 Chr.9: 127732596-127732596 on Build GRCh38
rs186889170 Chr.9: 127732657-127732657 on Build GRCh38
rs747261670 Chr.9: 127732638-127732638 on Build GRCh38
rs1003294939 Chr.9: 127732612-127732612 on Build GRCh38
rs750036702 Chr.9: 127732675-127732675 on Build GRCh38
rs552619025 Chr.9: 127732615-127732615 on Build GRCh38
rs755853444 Chr.9: 127732621-127732621 on Build GRCh38
rs753722516 Chr.9: 127732683-127732683 on Build GRCh38
rs774527500 Chr.9: 127732569-127732569 on Build GRCh38
rs763686261 Chr.9: 127732533-127732533 on Build GRCh38
rs745722125 Chr.9: 127732542-127732542 on Build GRCh38
rs756041438 Chr.9: 127732547-127732547 on Build GRCh38
rs867376298 Chr.9: 127732645-127732645 on Build GRCh38
rs748323312 Chr.9: 127732630-127732630 on Build GRCh38
rs374370419 Chr.9: 127732686-127732686 on Build GRCh38
rs771329946 Chr.9: 127732701-127732701 on Build GRCh38
rs778088015 Chr.9: 127732636-127732636 on Build GRCh38
rs756998924 Chr.9: 127732505-127732505 on Build GRCh38
rs781398751 Chr.9: 127732709-127732709 on Build GRCh38
rs766138297 Chr.9: 127732679-127732679 on Build GRCh38
rs201342330 Chr.9: 127732588-127732588 on Build GRCh38
rs767036531 Chr.9: 127732670-127732670 on Build GRCh38
rs367759175 Chr.9: 127732583-127732583 on Build GRCh38
rs773757831 Chr.9: 127732522-127732522 on Build GRCh38
rs778033031 Chr.9: 127732688-127732688 on Build GRCh38
rs778839588 Chr.9: 127732685-127732685 on Build GRCh38
rs538571049 Chr.9: 127732500-127732500 on Build GRCh38
rs990423836 Chr.9: 127732499-127732499 on Build GRCh38
rs200744648 Chr.9: 127732599-127732599 on Build GRCh38
rs515182 Chr.9: 127732532-127732532 on Build GRCh38
rs767055587 Chr.9: 127732526-127732526 on Build GRCh38
rs528717082 Chr.9: 127732635-127732635 on Build GRCh38
rs368374441 Chr.9: 127732585-127732585 on Build GRCh38
rs371662040 Chr.9: 127732536-127732536 on Build GRCh38
rs758507010 Chr.9: 127732687-127732687 on Build GRCh38
rs201869689 Chr.9: 127732639-127732639 on Build GRCh38
rs868147285 Chr.9: 127732597-127732597 on Build GRCh38
rs538066 Chr.9: 127732678-127732678 on Build GRCh38
rs925747863 Chr.9: 127732507-127732507 on Build GRCh38
rs753684849 Chr.9: 127732628-127732628 on Build GRCh38
rs767739421 Chr.9: 127732520-127732520 on Build GRCh38
rs371706865 Chr.9: 127732658-127732658 on Build GRCh38

View Full Product Details