Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 152126265-152126532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STAM2
Entrez Gene ID
10254
Gene Name
signal transducing adaptor molecule 2
Gene Aliases
Hbp, STAM2A, STAM2B
UniGene
Hs.17200
Gene Chromosome Location
Chr.2: 152116801-152175992 on Build GRCh38
Transcripts
NM_005843.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7274093 c.1111T>C p.Y371H Chr.2: 152126294-152126294 on Build GRCh38
6262184 c.1124C>A p.S375* Chr.2: 152126281-152126281 on Build GRCh38
6397608 c.1054G>T p.V352F Chr.2: 152126351-152126351 on Build GRCh38
5539773 c.1072C>T p.L358L Chr.2: 152126333-152126333 on Build GRCh38
441262 c.1095G>A p.V365V Chr.2: 152126310-152126310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199899749 Chr.2: 152126384-152126384 on Build GRCh38
rs771071364 Chr.2: 152126391-152126391 on Build GRCh38
rs750134293 Chr.2: 152126313-152126313 on Build GRCh38
rs1032428000 Chr.2: 152126348-152126348 on Build GRCh38
rs555939907 Chr.2: 152126385-152126385 on Build GRCh38
rs760601294 Chr.2: 152126292-152126292 on Build GRCh38
rs779676100 Chr.2: 152126325-152126325 on Build GRCh38
rs372669457 Chr.2: 152126401-152126401 on Build GRCh38
rs1041100780 Chr.2: 152126441-152126441 on Build GRCh38
rs908292000 Chr.2: 152126420-152126420 on Build GRCh38
rs911492070 Chr.2: 152126422-152126422 on Build GRCh38
rs939666687 Chr.2: 152126432-152126432 on Build GRCh38
rs755446266 Chr.2: 152126355-152126355 on Build GRCh38
rs965633355 Chr.2: 152126472-152126472 on Build GRCh38
rs371684032 Chr.2: 152126317-152126317 on Build GRCh38
rs751045492 Chr.2: 152126343-152126343 on Build GRCh38
rs764129439 Chr.2: 152126297-152126297 on Build GRCh38
rs781688227 Chr.2: 152126362-152126362 on Build GRCh38
rs556422738 Chr.2: 152126290-152126290 on Build GRCh38
rs932544383 Chr.2: 152126485-152126485 on Build GRCh38
rs375749442 Chr.2: 152126496-152126496 on Build GRCh38
rs922333176 Chr.2: 152126477-152126477 on Build GRCh38
rs770183641 Chr.2: 152126377-152126377 on Build GRCh38
rs759561723 Chr.2: 152126411-152126411 on Build GRCh38
rs748464839 Chr.2: 152126364-152126364 on Build GRCh38
rs760676879 Chr.2: 152126379-152126392 on Build GRCh38
rs139681962 Chr.2: 152126300-152126300 on Build GRCh38
rs764965275 Chr.2: 152126303-152126303 on Build GRCh38
rs374974704 Chr.2: 152126412-152126412 on Build GRCh38

View Full Product Details