Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 31728952-31729181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMN1
Entrez Gene ID
196394
Gene Name
antagonist of mitotic exit network 1 homolog
Gene Aliases
-
UniGene
Hs.585084
Gene Chromosome Location
Chr.12: 31671137-31729178 on Build GRCh38
Transcripts
NM_001113402.1 NM_001278411.1 NM_001278412.1 NR_103522.1 NR_103523.1 XM_011520604.2 XM_017018963.1 XM_017018965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3704073 c.21C>T p.V7V Chr.12: 31728988-31728988 on Build GRCh38
6234308 c.38+10C>A p.? Chr.12: 31728961-31728961 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs575158084 Chr.12: 31729070-31729070 on Build GRCh38
rs566143733 Chr.12: 31729000-31729000 on Build GRCh38
rs762331161 Chr.12: 31729046-31729046 on Build GRCh38
rs968784249 Chr.12: 31729002-31729002 on Build GRCh38
rs1039227467 Chr.12: 31728998-31728998 on Build GRCh38
rs774824428 Chr.12: 31729021-31729021 on Build GRCh38
rs905101960 Chr.12: 31729096-31729096 on Build GRCh38
rs114266500 Chr.12: 31729094-31729094 on Build GRCh38
rs769372231 Chr.12: 31729018-31729018 on Build GRCh38
rs891435599 Chr.12: 31729045-31729045 on Build GRCh38
rs1054069201 Chr.12: 31729043-31729043 on Build GRCh38
rs552636079 Chr.12: 31728996-31728996 on Build GRCh38
rs1045252937 Chr.12: 31728989-31728989 on Build GRCh38
rs139742682 Chr.12: 31729050-31729050 on Build GRCh38
rs918619668 Chr.12: 31729104-31729104 on Build GRCh38
rs886889363 Chr.12: 31729138-31729138 on Build GRCh38
rs1048516892 Chr.12: 31729137-31729137 on Build GRCh38
rs951341827 Chr.12: 31729134-31729134 on Build GRCh38
rs759648671 Chr.12: 31729009-31729010 on Build GRCh38
rs11551369 Chr.12: 31729072-31729072 on Build GRCh38
rs774129478 Chr.12: 31729037-31729037 on Build GRCh38
rs538794860 Chr.12: 31728987-31728987 on Build GRCh38
rs556230960 Chr.12: 31729073-31729073 on Build GRCh38
rs1034563920 Chr.12: 31729089-31729089 on Build GRCh38
rs931032922 Chr.12: 31729131-31729131 on Build GRCh38
rs767926699 Chr.12: 31729048-31729048 on Build GRCh38
rs535179290 Chr.12: 31729036-31729036 on Build GRCh38
rs992897604 Chr.12: 31729052-31729052 on Build GRCh38
rs1004380580 Chr.12: 31729159-31729159 on Build GRCh38
rs544824735 Chr.12: 31729155-31729155 on Build GRCh38

View Full Product Details