Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 18672515-18672772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL5B
Entrez Gene ID
221079
Gene Name
ADP ribosylation factor like GTPase 5B
Gene Aliases
ARL8
UniGene
Hs.25362
Gene Chromosome Location
Chr.10: 18659335-18681639 on Build GRCh38
Transcripts
NM_178815.3 XM_005252400.1 XM_005252401.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5809124 c.271G>T p.V91F Chr.10: 18672637-18672637 on Build GRCh38
6840659 c.317T>C p.L106S Chr.10: 18672683-18672683 on Build GRCh38
4839315 c.313G>T p.E105* Chr.10: 18672679-18672679 on Build GRCh38
4833573 c.283G>T p.D95Y Chr.10: 18672649-18672649 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756404404 Chr.10: 18672688-18672688 on Build GRCh38
rs754014770 Chr.10: 18672721-18672721 on Build GRCh38
rs761294822 Chr.10: 18672684-18672684 on Build GRCh38
rs202027107 Chr.10: 18672746-18672746 on Build GRCh38
rs932377343 Chr.10: 18672564-18672564 on Build GRCh38
rs750783693 Chr.10: 18672678-18672678 on Build GRCh38
rs776265443 Chr.10: 18672603-18672603 on Build GRCh38
rs367724087 Chr.10: 18672565-18672565 on Build GRCh38
rs754946345 Chr.10: 18672731-18672731 on Build GRCh38
rs774945318 Chr.10: 18672624-18672624 on Build GRCh38
rs199917924 Chr.10: 18672600-18672600 on Build GRCh38
rs757824606 Chr.10: 18672599-18672603 on Build GRCh38
rs940969224 Chr.10: 18672615-18672615 on Build GRCh38
rs1036700453 Chr.10: 18672677-18672677 on Build GRCh38
rs922491642 Chr.10: 18672560-18672560 on Build GRCh38
rs976527936 Chr.10: 18672556-18672556 on Build GRCh38
rs11015531 Chr.10: 18672621-18672621 on Build GRCh38
rs566934471 Chr.10: 18672588-18672588 on Build GRCh38
rs778834601 Chr.10: 18672742-18672742 on Build GRCh38
rs931241286 Chr.10: 18672739-18672739 on Build GRCh38
rs767949340 Chr.10: 18672655-18672655 on Build GRCh38
rs561561053 Chr.10: 18672553-18672553 on Build GRCh38
rs538867066 Chr.10: 18672667-18672667 on Build GRCh38
rs750847552 Chr.10: 18672728-18672729 on Build GRCh38
rs766495533 Chr.10: 18672710-18672710 on Build GRCh38
rs758966581 Chr.10: 18672609-18672609 on Build GRCh38
rs770470297 Chr.10: 18672602-18672602 on Build GRCh38
rs773802951 Chr.10: 18672570-18672570 on Build GRCh38
rs751389635 Chr.10: 18672618-18672618 on Build GRCh38
rs369433772 Chr.10: 18672612-18672612 on Build GRCh38
rs781531395 Chr.10: 18672632-18672634 on Build GRCh38
rs751554273 Chr.10: 18672572-18672573 on Build GRCh38
rs919729545 Chr.10: 18672681-18672681 on Build GRCh38
rs1014527274 Chr.10: 18672725-18672725 on Build GRCh38
rs771806741 Chr.10: 18672575-18672575 on Build GRCh38

View Full Product Details