Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 12919430-12919683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLR8
Entrez Gene ID
51311
Gene Name
toll like receptor 8
Gene Aliases
CD288
UniGene
Hs.660543
Gene Chromosome Location
Chr.X: 12906620-12923169 on Build GRCh38
Transcripts
NM_016610.3 NM_138636.5 XM_011545529.1 XM_011545530.2 NR_030727.1

Gene Symbol
TLR8-AS1
Entrez Gene ID
349408
Gene Name
TLR8 antisense RNA 1
Gene Aliases
GS1-324M7.6
UniGene
Hs.685035
Gene Chromosome Location
Chr.X: 12902817-12943300 on Build GRCh38
Transcripts
NM_016610.3 NM_138636.5 XM_011545529.1 XM_011545530.2 NR_030727.1

SNP Detail

SNP ID Chromosome Location
rs768640826 Chr.X: 12919497-12919497 on Build GRCh38
rs5744079 Chr.X: 12919601-12919601 on Build GRCh38
rs749440648 Chr.X: 12919526-12919526 on Build GRCh38
rs376689229 Chr.X: 12919512-12919512 on Build GRCh38
rs761066050 Chr.X: 12919565-12919565 on Build GRCh38
rs779613268 Chr.X: 12919638-12919638 on Build GRCh38
rs754297567 Chr.X: 12919585-12919585 on Build GRCh38
rs776057581 Chr.X: 12919600-12919600 on Build GRCh38
rs767811815 Chr.X: 12919549-12919549 on Build GRCh38
rs183801068 Chr.X: 12919532-12919532 on Build GRCh38
rs776006574 Chr.X: 12919554-12919554 on Build GRCh38
rs746819165 Chr.X: 12919640-12919640 on Build GRCh38
rs779846666 Chr.X: 12919457-12919457 on Build GRCh38
rs759806873 Chr.X: 12919535-12919535 on Build GRCh38
rs746766277 Chr.X: 12919490-12919490 on Build GRCh38
rs756834432 Chr.X: 12919589-12919589 on Build GRCh38
rs976964102 Chr.X: 12919636-12919636 on Build GRCh38
rs934606168 Chr.X: 12919616-12919616 on Build GRCh38
rs754712471 Chr.X: 12919644-12919644 on Build GRCh38
rs932494695 Chr.X: 12919645-12919645 on Build GRCh38
rs752683730 Chr.X: 12919567-12919567 on Build GRCh38
rs148046494 Chr.X: 12919637-12919637 on Build GRCh38
rs762362421 Chr.X: 12919519-12919519 on Build GRCh38
rs376034872 Chr.X: 12919602-12919602 on Build GRCh38

View Full Product Details