Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92134237-92134494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF2
Entrez Gene ID
53981
Gene Name
cleavage and polyadenylation specific factor 2
Gene Aliases
CPSF100
UniGene
Hs.657632
Gene Chromosome Location
Chr.14: 92121954-92164199 on Build GRCh38
Transcripts
NM_001322270.1 NM_001322271.1 NM_001322272.1 NM_017437.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6076559 c.314G>A p.R105Q Chr.14: 92134254-92134254 on Build GRCh38
4578064 c.365C>A p.A122D Chr.14: 92134305-92134305 on Build GRCh38
1629432 c.392T>A p.F131Y Chr.14: 92134332-92134332 on Build GRCh38
1202198 c.328G>T p.D110Y Chr.14: 92134268-92134268 on Build GRCh38
2251863 c.375A>C p.K125N Chr.14: 92134315-92134315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2295726 Chr.14: 92134454-92134454 on Build GRCh38
rs760567119 Chr.14: 92134356-92134356 on Build GRCh38
rs772967956 Chr.14: 92134345-92134345 on Build GRCh38
rs371087877 Chr.14: 92134297-92134297 on Build GRCh38
rs1033577194 Chr.14: 92134424-92134424 on Build GRCh38
rs200651259 Chr.14: 92134276-92134276 on Build GRCh38
rs758152496 Chr.14: 92134285-92134285 on Build GRCh38
rs2295727 Chr.14: 92134441-92134441 on Build GRCh38
rs758079128 Chr.14: 92134402-92134402 on Build GRCh38
rs766332624 Chr.14: 92134381-92134381 on Build GRCh38
rs34963129 Chr.14: 92134347-92134348 on Build GRCh38
rs558577129 Chr.14: 92134394-92134394 on Build GRCh38
rs890061655 Chr.14: 92134277-92134277 on Build GRCh38
rs955887280 Chr.14: 92134379-92134379 on Build GRCh38
rs967154472 Chr.14: 92134455-92134455 on Build GRCh38
rs572034512 Chr.14: 92134396-92134396 on Build GRCh38
rs769969271 Chr.14: 92134279-92134279 on Build GRCh38
rs763119466 Chr.14: 92134301-92134301 on Build GRCh38
rs767322661 Chr.14: 92134327-92134327 on Build GRCh38
rs752433588 Chr.14: 92134400-92134400 on Build GRCh38
rs959294308 Chr.14: 92134438-92134446 on Build GRCh38
rs1006086815 Chr.14: 92134418-92134420 on Build GRCh38
rs548849313 Chr.14: 92134422-92134422 on Build GRCh38
rs540853686 Chr.14: 92134417-92134417 on Build GRCh38

View Full Product Details