Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 69719400-69719649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL1
Entrez Gene ID
57626
Gene Name
kelch like family member 1
Gene Aliases
MRP2
UniGene
Hs.508201
Gene Chromosome Location
Chr.13: 69700593-70108493 on Build GRCh38
Transcripts
NM_001286725.1 NM_020866.2 XM_017020678.1 XM_017020679.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7374130 c.1853A>C p.E618A Chr.13: 69719531-69719531 on Build GRCh38
5809773 c.1975G>T p.A659S Chr.13: 69719409-69719409 on Build GRCh38
5439084 c.1911A>T p.R637S Chr.13: 69719473-69719473 on Build GRCh38
1706832 c.1984C>T p.H662Y Chr.13: 69719400-69719400 on Build GRCh38
5823769 c.1938C>T p.D646D Chr.13: 69719446-69719446 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759184108 Chr.13: 69719511-69719511 on Build GRCh38
rs751899459 Chr.13: 69719527-69719527 on Build GRCh38
rs781599073 Chr.13: 69719446-69719446 on Build GRCh38
rs756108438 Chr.13: 69719456-69719456 on Build GRCh38
rs779224372 Chr.13: 69719581-69719581 on Build GRCh38
rs762034599 Chr.13: 69719530-69719530 on Build GRCh38
rs201683214 Chr.13: 69719458-69719458 on Build GRCh38
rs11336815 Chr.13: 69719584-69719584 on Build GRCh38
rs764909620 Chr.13: 69719525-69719525 on Build GRCh38
rs146957152 Chr.13: 69719494-69719494 on Build GRCh38
rs773727834 Chr.13: 69719614-69719616 on Build GRCh38
rs750832204 Chr.13: 69719561-69719561 on Build GRCh38
rs372198907 Chr.13: 69719464-69719464 on Build GRCh38
rs376662373 Chr.13: 69719463-69719463 on Build GRCh38
rs141275075 Chr.13: 69719490-69719490 on Build GRCh38
rs756660309 Chr.13: 69719567-69719567 on Build GRCh38
rs758059208 Chr.13: 69719608-69719608 on Build GRCh38
rs765942820 Chr.13: 69719425-69719425 on Build GRCh38
rs147995833 Chr.13: 69719614-69719614 on Build GRCh38
rs530644820 Chr.13: 69719493-69719493 on Build GRCh38
rs375572650 Chr.13: 69719444-69719444 on Build GRCh38
rs201140611 Chr.13: 69719445-69719445 on Build GRCh38
rs531126589 Chr.13: 69719450-69719450 on Build GRCh38
rs771918362 Chr.13: 69719479-69719479 on Build GRCh38
rs371876461 Chr.13: 69719440-69719440 on Build GRCh38
rs753364231 Chr.13: 69719436-69719436 on Build GRCh38
rs746905688 Chr.13: 69719619-69719619 on Build GRCh38
rs748410496 Chr.13: 69719604-69719604 on Build GRCh38
rs903420817 Chr.13: 69719462-69719462 on Build GRCh38
rs398098952 Chr.13: 69719588-69719588 on Build GRCh38
rs767957839 Chr.13: 69719535-69719535 on Build GRCh38
rs764429311 Chr.13: 69719441-69719441 on Build GRCh38
rs865978178 Chr.13: 69719618-69719618 on Build GRCh38
rs914755690 Chr.13: 69719428-69719428 on Build GRCh38
rs766360091 Chr.13: 69719578-69719578 on Build GRCh38
rs778850414 Chr.13: 69719468-69719468 on Build GRCh38
rs143518222 Chr.13: 69719506-69719506 on Build GRCh38

View Full Product Details