Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159020334-159020593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFI16
Entrez Gene ID
3428
Gene Name
interferon gamma inducible protein 16
Gene Aliases
IFNGIP1, PYHIN2
UniGene
Hs.380250
Gene Chromosome Location
Chr.1: 158999968-159055155 on Build GRCh38
Transcripts
NM_001206567.1 NM_005531.2 XM_005245127.3 XM_006711290.1 XM_017001149.1 XM_017001150.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3418195 c.1003G>A p.E335K Chr.1: 159020371-159020371 on Build GRCh38
5799407 c.1133T>A p.L378Q Chr.1: 159020501-159020501 on Build GRCh38
5799406 c.1133T>A p.L378Q Chr.1: 159020501-159020501 on Build GRCh38
3369362 c.1103G>A p.R368Q Chr.1: 159020471-159020471 on Build GRCh38
4819870 c.1134G>C p.L378L Chr.1: 159020502-159020502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745565983 Chr.1: 159020429-159020429 on Build GRCh38
rs1020454409 Chr.1: 159020479-159020479 on Build GRCh38
rs776250741 Chr.1: 159020382-159020382 on Build GRCh38
rs201766347 Chr.1: 159020467-159020467 on Build GRCh38
rs753994441 Chr.1: 159020458-159020458 on Build GRCh38
rs769517369 Chr.1: 159020400-159020400 on Build GRCh38
rs754982221 Chr.1: 159020552-159020552 on Build GRCh38
rs759905608 Chr.1: 159020426-159020426 on Build GRCh38
rs367941160 Chr.1: 159020433-159020433 on Build GRCh38
rs758261163 Chr.1: 159020542-159020542 on Build GRCh38
rs750172788 Chr.1: 159020501-159020501 on Build GRCh38
rs751516237 Chr.1: 159020546-159020546 on Build GRCh38
rs780142234 Chr.1: 159020368-159020368 on Build GRCh38
rs199911779 Chr.1: 159020460-159020460 on Build GRCh38
rs1001431500 Chr.1: 159020394-159020394 on Build GRCh38
rs200970346 Chr.1: 159020544-159020544 on Build GRCh38
rs527791605 Chr.1: 159020452-159020452 on Build GRCh38
rs774647644 Chr.1: 159020418-159020418 on Build GRCh38
rs1057024 Chr.1: 159020457-159020457 on Build GRCh38
rs775974310 Chr.1: 159020444-159020444 on Build GRCh38
rs867123527 Chr.1: 159020363-159020363 on Build GRCh38
rs758365324 Chr.1: 159020365-159020365 on Build GRCh38
rs75796001 Chr.1: 159020371-159020371 on Build GRCh38
rs199556510 Chr.1: 159020370-159020370 on Build GRCh38
rs371364809 Chr.1: 159020411-159020411 on Build GRCh38
rs772502920 Chr.1: 159020427-159020427 on Build GRCh38
rs377636508 Chr.1: 159020385-159020385 on Build GRCh38

View Full Product Details