Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 42234287-42234534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GSK3A
Entrez Gene ID
2931
Gene Name
glycogen synthase kinase 3 alpha
Gene Aliases
-
UniGene
Hs.466828
Gene Chromosome Location
Chr.19: 42230186-42243330 on Build GRCh38
Transcripts
NM_019884.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3286990 c.903C>T p.I301I Chr.19: 42234354-42234354 on Build GRCh38
3286992 c.814C>T p.R272* Chr.19: 42234443-42234443 on Build GRCh38
6208958 c.857G>A p.R286Q Chr.19: 42234400-42234400 on Build GRCh38
997280 c.850T>C p.Y284H Chr.19: 42234407-42234407 on Build GRCh38
5418712 c.891C>T p.Y297Y Chr.19: 42234366-42234366 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1035974110 Chr.19: 42234409-42234409 on Build GRCh38
rs753711736 Chr.19: 42234456-42234456 on Build GRCh38
rs1002040368 Chr.19: 42234499-42234499 on Build GRCh38
rs770210270 Chr.19: 42234503-42234503 on Build GRCh38
rs759105475 Chr.19: 42234410-42234410 on Build GRCh38
rs774840736 Chr.19: 42234442-42234442 on Build GRCh38
rs745584114 Chr.19: 42234510-42234510 on Build GRCh38
rs373598275 Chr.19: 42234343-42234343 on Build GRCh38
rs747597403 Chr.19: 42234342-42234342 on Build GRCh38
rs760709783 Chr.19: 42234451-42234451 on Build GRCh38
rs779230727 Chr.19: 42234325-42234325 on Build GRCh38
rs780218834 Chr.19: 42234481-42234481 on Build GRCh38
rs370945979 Chr.19: 42234488-42234488 on Build GRCh38
rs750744981 Chr.19: 42234480-42234480 on Build GRCh38
rs976079367 Chr.19: 42234319-42234319 on Build GRCh38
rs767113418 Chr.19: 42234429-42234429 on Build GRCh38
rs36103293 Chr.19: 42234401-42234401 on Build GRCh38
rs547256347 Chr.19: 42234453-42234453 on Build GRCh38
rs748515703 Chr.19: 42234497-42234497 on Build GRCh38
rs753535275 Chr.19: 42234325-42234325 on Build GRCh38
rs565510503 Chr.19: 42234479-42234479 on Build GRCh38
rs781620457 Chr.19: 42234490-42234490 on Build GRCh38
rs747018124 Chr.19: 42234487-42234487 on Build GRCh38
rs905036035 Chr.19: 42234495-42234495 on Build GRCh38
rs757588530 Chr.19: 42234317-42234317 on Build GRCh38
rs757063381 Chr.19: 42234473-42234473 on Build GRCh38
rs745963375 Chr.19: 42234328-42234328 on Build GRCh38
rs770857449 Chr.19: 42234330-42234330 on Build GRCh38
rs772126918 Chr.19: 42234329-42234329 on Build GRCh38
rs777008705 Chr.19: 42234346-42234346 on Build GRCh38
rs369723953 Chr.19: 42234311-42234311 on Build GRCh38
rs762115653 Chr.19: 42234354-42234354 on Build GRCh38
rs538863467 Chr.19: 42234314-42234314 on Build GRCh38
rs376493455 Chr.19: 42234357-42234357 on Build GRCh38
rs773553059 Chr.19: 42234506-42234506 on Build GRCh38
rs981266433 Chr.19: 42234401-42234401 on Build GRCh38
rs754589092 Chr.19: 42234506-42234507 on Build GRCh38
rs777675402 Chr.19: 42234312-42234312 on Build GRCh38

View Full Product Details