Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 85025893-85026162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCOLN3
Entrez Gene ID
55283
Gene Name
mucolipin 3
Gene Aliases
TRP-ML3, TRPML3
UniGene
Hs.535239
Gene Chromosome Location
Chr.1: 85018082-85048902 on Build GRCh38
Transcripts
NM_001253693.1 NM_018298.10 XM_005271003.1 XM_006710750.1 XM_011541739.2 XM_011541740.2 XM_011541741.2 XM_011541742.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5892615 c.946-8C>T p.? Chr.1: 85026096-85026096 on Build GRCh38
2157476 c.1061T>C p.I354T Chr.1: 85025973-85025973 on Build GRCh38
3985256 c.1017T>C p.N339N Chr.1: 85026017-85026017 on Build GRCh38
4165312 c.1085T>C p.I362T Chr.1: 85025949-85025949 on Build GRCh38
1662725 c.1093A>T p.K365* Chr.1: 85025941-85025941 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550757294 Chr.1: 85026004-85026004 on Build GRCh38
rs778293236 Chr.1: 85026008-85026008 on Build GRCh38
rs922922675 Chr.1: 85025940-85025940 on Build GRCh38
rs201828020 Chr.1: 85025923-85025923 on Build GRCh38
rs751354499 Chr.1: 85025928-85025928 on Build GRCh38
rs761066830 Chr.1: 85026036-85026036 on Build GRCh38
rs945782332 Chr.1: 85025926-85025926 on Build GRCh38
rs781180261 Chr.1: 85025929-85025929 on Build GRCh38
rs111585611 Chr.1: 85026103-85026103 on Build GRCh38
rs757142286 Chr.1: 85025928-85025928 on Build GRCh38
rs754967173 Chr.1: 85026097-85026097 on Build GRCh38
rs752781847 Chr.1: 85026079-85026079 on Build GRCh38
rs1040727519 Chr.1: 85025973-85025973 on Build GRCh38
rs111729386 Chr.1: 85026041-85026041 on Build GRCh38
rs760695983 Chr.1: 85026063-85026063 on Build GRCh38
rs1050256005 Chr.1: 85026092-85026092 on Build GRCh38
rs771314165 Chr.1: 85026049-85026049 on Build GRCh38
rs773685792 Chr.1: 85026022-85026022 on Build GRCh38
rs768299294 Chr.1: 85026005-85026005 on Build GRCh38
rs377394905 Chr.1: 85026116-85026116 on Build GRCh38
rs745711354 Chr.1: 85025938-85025938 on Build GRCh38
rs190703120 Chr.1: 85026107-85026107 on Build GRCh38
rs750419923 Chr.1: 85026098-85026098 on Build GRCh38
rs765430597 Chr.1: 85026070-85026070 on Build GRCh38
rs1050773817 Chr.1: 85026020-85026020 on Build GRCh38
rs747660031 Chr.1: 85026129-85026129 on Build GRCh38
rs747689272 Chr.1: 85026011-85026011 on Build GRCh38
rs945996922 Chr.1: 85025955-85025955 on Build GRCh38
rs369107433 Chr.1: 85025971-85025971 on Build GRCh38
rs755186272 Chr.1: 85025922-85025922 on Build GRCh38
rs754654907 Chr.1: 85026112-85026112 on Build GRCh38
rs772513559 Chr.1: 85026018-85026018 on Build GRCh38
rs149878300 Chr.1: 85026065-85026065 on Build GRCh38
rs755945050 Chr.1: 85025970-85025970 on Build GRCh38
rs748972987 Chr.1: 85026002-85026002 on Build GRCh38
rs767607198 Chr.1: 85026094-85026094 on Build GRCh38
rs141043298 Chr.1: 85026080-85026080 on Build GRCh38
rs776930920 Chr.1: 85026060-85026060 on Build GRCh38

View Full Product Details