Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 24419457-24419714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NIPAL3
Entrez Gene ID
57185
Gene Name
NIPA like domain containing 3
Gene Aliases
DJ462O23.2, NPAL3
UniGene
Hs.523442
Gene Chromosome Location
Chr.1: 24413360-24472983 on Build GRCh38
Transcripts
NM_001322854.1 NM_001322855.1 NM_001322856.1 NM_001322857.1 NM_001322858.1 NM_001322859.1 NM_001322860.1 NM_001322861.1 NM_001322862.1 NM_001322863.1 NM_001322864.1 NM_001322865.1 NM_001322866.1 NM_001322868.1 NM_020448.4 NR_136506.1 NR_136507.1 NR_136508.1 XM_011541808.2 XM_017001868.1 XM_017001869.1 XM_017001870.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6354204 c.90C>T p.Y30Y Chr.1: 24419637-24419637 on Build GRCh38
5487660 c.1A>G p.M1V Chr.1: 24419548-24419548 on Build GRCh38
7379626 c.19G>A p.A7T Chr.1: 24419566-24419566 on Build GRCh38
5421095 c.42G>A p.L14L Chr.1: 24419589-24419589 on Build GRCh38
5421096 c.42G>A p.L14L Chr.1: 24419589-24419589 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768868473 Chr.1: 24419480-24419480 on Build GRCh38
rs757297751 Chr.1: 24419529-24419529 on Build GRCh38
rs775065369 Chr.1: 24419604-24419604 on Build GRCh38
rs377426633 Chr.1: 24419632-24419632 on Build GRCh38
rs1054425143 Chr.1: 24419517-24419517 on Build GRCh38
rs185544511 Chr.1: 24419535-24419535 on Build GRCh38
rs1025136724 Chr.1: 24419525-24419525 on Build GRCh38
rs764651166 Chr.1: 24419508-24419508 on Build GRCh38
rs1000552928 Chr.1: 24419505-24419505 on Build GRCh38
rs779010032 Chr.1: 24419654-24419654 on Build GRCh38
rs954021128 Chr.1: 24419617-24419617 on Build GRCh38
rs200988381 Chr.1: 24419678-24419678 on Build GRCh38
rs761862441 Chr.1: 24419554-24419554 on Build GRCh38
rs912361961 Chr.1: 24419663-24419663 on Build GRCh38
rs1012818577 Chr.1: 24419524-24419524 on Build GRCh38
rs780306794 Chr.1: 24419631-24419631 on Build GRCh38
rs765763781 Chr.1: 24419614-24419614 on Build GRCh38
rs745372455 Chr.1: 24419656-24419656 on Build GRCh38
rs758976558 Chr.1: 24419582-24419582 on Build GRCh38
rs747916172 Chr.1: 24419545-24419545 on Build GRCh38
rs775179228 Chr.1: 24419672-24419672 on Build GRCh38
rs779014257 Chr.1: 24419533-24419533 on Build GRCh38
rs767142086 Chr.1: 24419593-24419593 on Build GRCh38
rs749131065 Chr.1: 24419638-24419638 on Build GRCh38
rs756451494 Chr.1: 24419498-24419498 on Build GRCh38
rs34807044 Chr.1: 24419553-24419553 on Build GRCh38
rs902146544 Chr.1: 24419486-24419486 on Build GRCh38
rs750435540 Chr.1: 24419619-24419619 on Build GRCh38
rs770018746 Chr.1: 24419628-24419630 on Build GRCh38
rs372415650 Chr.1: 24419534-24419534 on Build GRCh38
rs895424654 Chr.1: 24419519-24419519 on Build GRCh38
rs2294521 Chr.1: 24419637-24419637 on Build GRCh38
rs923734336 Chr.1: 24419685-24419685 on Build GRCh38
rs373605055 Chr.1: 24419620-24419620 on Build GRCh38
rs369128212 Chr.1: 24419613-24419613 on Build GRCh38
rs754410818 Chr.1: 24419514-24419514 on Build GRCh38
rs919823909 Chr.1: 24419592-24419592 on Build GRCh38
rs761097882 Chr.1: 24419608-24419608 on Build GRCh38
rs749529629 Chr.1: 24419649-24419649 on Build GRCh38
rs896205168 Chr.1: 24419653-24419653 on Build GRCh38
rs773570339 Chr.1: 24419658-24419658 on Build GRCh38
rs963684002 Chr.1: 24419557-24419557 on Build GRCh38
rs771274080 Chr.1: 24419667-24419667 on Build GRCh38
rs778104359 Chr.1: 24419631-24419633 on Build GRCh38
rs148391167 Chr.1: 24419566-24419566 on Build GRCh38
rs770929415 Chr.1: 24419645-24419645 on Build GRCh38
rs749273872 Chr.1: 24419565-24419565 on Build GRCh38
rs771551575 Chr.1: 24419664-24419664 on Build GRCh38
rs773962283 Chr.1: 24419577-24419577 on Build GRCh38
rs943861086 Chr.1: 24419665-24419665 on Build GRCh38
rs764482874 Chr.1: 24419610-24419610 on Build GRCh38
rs975521633 Chr.1: 24419495-24419495 on Build GRCh38

View Full Product Details