Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 93312979-93313252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYOF
Entrez Gene ID
26509
Gene Name
myoferlin
Gene Aliases
FER1L3
UniGene
Hs.602086
Gene Chromosome Location
Chr.10: 93306429-93482504 on Build GRCh38
Transcripts
NM_013451.3 NM_133337.2 XM_005269693.4 XM_005269694.4 XM_011539632.2 XM_017016068.1 XM_017016069.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1297508 c.5724C>T p.H1908H Chr.10: 93313185-93313185 on Build GRCh38
4501141 c.5817C>T p.S1939S Chr.10: 93313092-93313092 on Build GRCh38
6720882 c.5849G>A p.R1950H Chr.10: 93313030-93313030 on Build GRCh38
3686996 c.5746C>T p.P1916S Chr.10: 93313163-93313163 on Build GRCh38
1349851 c.5719C>T p.R1907C Chr.10: 93313190-93313190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773972612 Chr.10: 93313169-93313169 on Build GRCh38
rs772978810 Chr.10: 93313057-93313057 on Build GRCh38
rs910738694 Chr.10: 93313192-93313192 on Build GRCh38
rs199884146 Chr.10: 93313135-93313135 on Build GRCh38
rs750870337 Chr.10: 93313105-93313105 on Build GRCh38
rs535486923 Chr.10: 93313104-93313104 on Build GRCh38
rs1002789680 Chr.10: 93313063-93313063 on Build GRCh38
rs760083271 Chr.10: 93313190-93313191 on Build GRCh38
rs747282819 Chr.10: 93313134-93313134 on Build GRCh38
rs200024635 Chr.10: 93313061-93313061 on Build GRCh38
rs865971927 Chr.10: 93313210-93313210 on Build GRCh38
rs781234710 Chr.10: 93313136-93313136 on Build GRCh38
rs898897534 Chr.10: 93313183-93313183 on Build GRCh38
rs370100946 Chr.10: 93313080-93313080 on Build GRCh38
rs138208183 Chr.10: 93313190-93313190 on Build GRCh38
rs377745370 Chr.10: 93313049-93313049 on Build GRCh38
rs769804478 Chr.10: 93313056-93313056 on Build GRCh38
rs201029088 Chr.10: 93313041-93313041 on Build GRCh38
rs748256668 Chr.10: 93313227-93313227 on Build GRCh38
rs771355145 Chr.10: 93313106-93313107 on Build GRCh38
rs769422043 Chr.10: 93313147-93313147 on Build GRCh38
rs989872821 Chr.10: 93313038-93313038 on Build GRCh38
rs751672361 Chr.10: 93313215-93313215 on Build GRCh38
rs370918123 Chr.10: 93313182-93313182 on Build GRCh38
rs754966864 Chr.10: 93313218-93313218 on Build GRCh38
rs374263594 Chr.10: 93313189-93313189 on Build GRCh38
rs780095527 Chr.10: 93313131-93313131 on Build GRCh38
rs765916973 Chr.10: 93313072-93313072 on Build GRCh38
rs778193566 Chr.10: 93313019-93313019 on Build GRCh38
rs374294319 Chr.10: 93313031-93313031 on Build GRCh38
rs530974439 Chr.10: 93313029-93313029 on Build GRCh38
rs759301010 Chr.10: 93313171-93313171 on Build GRCh38
rs764412531 Chr.10: 93313096-93313096 on Build GRCh38
rs1035967522 Chr.10: 93313083-93313083 on Build GRCh38
rs764598880 Chr.10: 93313094-93313094 on Build GRCh38
rs373595278 Chr.10: 93313112-93313112 on Build GRCh38
rs1024135707 Chr.10: 93313015-93313015 on Build GRCh38
rs201400625 Chr.10: 93313028-93313028 on Build GRCh38
rs1051889912 Chr.10: 93313113-93313113 on Build GRCh38
rs192250227 Chr.10: 93313050-93313050 on Build GRCh38
rs758541223 Chr.10: 93313116-93313116 on Build GRCh38
rs776963827 Chr.10: 93313109-93313110 on Build GRCh38
rs373169973 Chr.10: 93313097-93313097 on Build GRCh38
rs770620728 Chr.10: 93313160-93313160 on Build GRCh38
rs787666 Chr.10: 93313149-93313149 on Build GRCh38
rs749230643 Chr.10: 93313155-93313155 on Build GRCh38
rs145908975 Chr.10: 93313035-93313035 on Build GRCh38
rs972046372 Chr.10: 93313178-93313178 on Build GRCh38
rs369620858 Chr.10: 93313030-93313030 on Build GRCh38
rs758881685 Chr.10: 93313199-93313199 on Build GRCh38
rs762252053 Chr.10: 93313185-93313185 on Build GRCh38
rs772089105 Chr.10: 93313034-93313034 on Build GRCh38
rs765698303 Chr.10: 93313201-93313202 on Build GRCh38
rs200526001 Chr.10: 93313145-93313145 on Build GRCh38
rs374472012 Chr.10: 93313220-93313220 on Build GRCh38
rs370747406 Chr.10: 93313200-93313200 on Build GRCh38

View Full Product Details