Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155197698-155197943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THBS3
Entrez Gene ID
7059
Gene Name
thrombospondin 3
Gene Aliases
TSP3
UniGene
Hs.169875
Gene Chromosome Location
Chr.1: 155195588-155209180 on Build GRCh38
Transcripts
NM_001252607.1 NM_001252608.1 NM_007112.4 NR_045553.1 XM_006711498.3 XM_011509926.2 XM_011509927.2 XM_011509928.2 XM_011509929.1 XM_011509930.2 XM_011509931.1 XM_011509932.2 XM_011509933.1 XM_011509934.1 XM_011509935.1 XM_011509936.1 XM_017002206.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1600913 c.2296G>T p.A766S Chr.1: 155197886-155197886 on Build GRCh38
3474880 c.2254-1G>A p.? Chr.1: 155197929-155197929 on Build GRCh38
424038 c.2292C>G p.G764G Chr.1: 155197890-155197890 on Build GRCh38
1600914 c.2282G>A p.S761N Chr.1: 155197900-155197900 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760111092 Chr.1: 155197800-155197803 on Build GRCh38
rs906234482 Chr.1: 155197898-155197898 on Build GRCh38
rs1040694186 Chr.1: 155197896-155197896 on Build GRCh38
rs1053640225 Chr.1: 155197744-155197744 on Build GRCh38
rs911465605 Chr.1: 155197804-155197805 on Build GRCh38
rs764503646 Chr.1: 155197834-155197834 on Build GRCh38
rs893703118 Chr.1: 155197773-155197773 on Build GRCh38
rs943729978 Chr.1: 155197821-155197821 on Build GRCh38
rs750994628 Chr.1: 155197869-155197869 on Build GRCh38
rs1009512112 Chr.1: 155197778-155197778 on Build GRCh38
rs74118415 Chr.1: 155197804-155197804 on Build GRCh38
rs754650696 Chr.1: 155197909-155197909 on Build GRCh38
rs769646561 Chr.1: 155197923-155197923 on Build GRCh38
rs370777859 Chr.1: 155197845-155197845 on Build GRCh38
rs1042076241 Chr.1: 155197781-155197781 on Build GRCh38
rs778592737 Chr.1: 155197896-155197901 on Build GRCh38
rs760987625 Chr.1: 155197831-155197831 on Build GRCh38
rs374515100 Chr.1: 155197863-155197863 on Build GRCh38
rs866488441 Chr.1: 155197788-155197788 on Build GRCh38
rs932454244 Chr.1: 155197735-155197735 on Build GRCh38
rs376544999 Chr.1: 155197864-155197864 on Build GRCh38
rs751137112 Chr.1: 155197902-155197902 on Build GRCh38
rs548609928 Chr.1: 155197810-155197810 on Build GRCh38
rs140640141 Chr.1: 155197916-155197916 on Build GRCh38
rs757785511 Chr.1: 155197871-155197871 on Build GRCh38
rs569681720 Chr.1: 155197780-155197780 on Build GRCh38
rs765833515 Chr.1: 155197865-155197865 on Build GRCh38
rs779616472 Chr.1: 155197876-155197876 on Build GRCh38
rs931568205 Chr.1: 155197798-155197798 on Build GRCh38
rs760791847 Chr.1: 155197745-155197745 on Build GRCh38

View Full Product Details