Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77422698-77422929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOXRED1
Entrez Gene ID
122945
Gene Name
NADP dependent oxidoreductase domain containing 1
Gene Aliases
C14orf148
UniGene
Hs.97556
Gene Chromosome Location
Chr.14: 77394021-77426013 on Build GRCh38
Transcripts
NM_001113475.2 XM_005267330.3 XM_011536427.2 XM_011536428.2 XM_011536429.2 XM_017020969.1 XM_017020970.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
319011 c.86G>C p.R29P Chr.14: 77422804-77422804 on Build GRCh38
958163 c.100A>G p.M34V Chr.14: 77422790-77422790 on Build GRCh38
6231082 c.15G>T p.Q5H Chr.14: 77422875-77422875 on Build GRCh38
7380117 c.19C>G p.L7V Chr.14: 77422871-77422871 on Build GRCh38
2139398 c.115G>A p.A39T Chr.14: 77422775-77422775 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762054982 Chr.14: 77422737-77422737 on Build GRCh38
rs909640599 Chr.14: 77422859-77422859 on Build GRCh38
rs972826520 Chr.14: 77422897-77422897 on Build GRCh38
rs772828730 Chr.14: 77422891-77422891 on Build GRCh38
rs148712624 Chr.14: 77422728-77422728 on Build GRCh38
rs190283547 Chr.14: 77422729-77422729 on Build GRCh38
rs367617483 Chr.14: 77422749-77422749 on Build GRCh38
rs371769717 Chr.14: 77422788-77422788 on Build GRCh38
rs781587634 Chr.14: 77422777-77422777 on Build GRCh38
rs866280108 Chr.14: 77422774-77422774 on Build GRCh38
rs776369560 Chr.14: 77422893-77422893 on Build GRCh38
rs983877611 Chr.14: 77422830-77422830 on Build GRCh38
rs780712348 Chr.14: 77422828-77422828 on Build GRCh38
rs146724025 Chr.14: 77422805-77422805 on Build GRCh38
rs375219656 Chr.14: 77422804-77422804 on Build GRCh38
rs769995887 Chr.14: 77422886-77422886 on Build GRCh38
rs765446270 Chr.14: 77422829-77422829 on Build GRCh38
rs757759337 Chr.14: 77422875-77422875 on Build GRCh38
rs976595560 Chr.14: 77422800-77422800 on Build GRCh38
rs781501210 Chr.14: 77422883-77422883 on Build GRCh38
rs1020842258 Chr.14: 77422784-77422784 on Build GRCh38
rs773352865 Chr.14: 77422801-77422801 on Build GRCh38
rs953791156 Chr.14: 77422820-77422820 on Build GRCh38
rs777823768 Chr.14: 77422850-77422850 on Build GRCh38
rs771750858 Chr.14: 77422889-77422889 on Build GRCh38
rs748262510 Chr.14: 77422727-77422727 on Build GRCh38
rs752044084 Chr.14: 77422852-77422852 on Build GRCh38
rs774106739 Chr.14: 77422790-77422790 on Build GRCh38
rs1009990862 Chr.14: 77422781-77422781 on Build GRCh38
rs778766698 Chr.14: 77422887-77422887 on Build GRCh38
rs758528801 Chr.14: 77422845-77422845 on Build GRCh38
rs371990284 Chr.14: 77422892-77422892 on Build GRCh38
rs761422368 Chr.14: 77422798-77422798 on Build GRCh38
rs1025944560 Chr.14: 77422856-77422856 on Build GRCh38
rs763306225 Chr.14: 77422745-77422747 on Build GRCh38
rs369588295 Chr.14: 77422822-77422822 on Build GRCh38
rs771696130 Chr.14: 77422799-77422799 on Build GRCh38
rs906030477 Chr.14: 77422770-77422770 on Build GRCh38
rs746275190 Chr.14: 77422885-77422885 on Build GRCh38
rs753693791 Chr.14: 77422806-77422806 on Build GRCh38
rs748941333 Chr.14: 77422785-77422785 on Build GRCh38
rs747799145 Chr.14: 77422888-77422888 on Build GRCh38
rs752856096 Chr.14: 77422837-77422837 on Build GRCh38

View Full Product Details