Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 139261408-139261645 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UBN2
Entrez Gene ID
254048
Gene Name
ubinuclein 2
Gene Aliases
-
UniGene
Hs.153458
Gene Chromosome Location
Chr.7: 139229565-139329421 on Build GRCh38
Transcripts
NM_173569.3 XM_005250249.3 XM_006715916.3 XM_011516002.2 XM_011516003.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3762421 c.1271C>T p.S424L Chr.7: 139261617-139261617 on Build GRCh38
387831 c.849G>T p.G283G Chr.7: 139261444-139261444 on Build GRCh38
6481192 c.1275G>T p.G425G Chr.7: 139261621-139261621 on Build GRCh38
5067313 c.1022_1023insG p.E344fs*1 Chr.7: 139261617-139261618 on Build GRCh38
6471673 c.1153G>T p.V385F Chr.7: 139261499-139261499 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs75136253 Chr.7: 139261550-139261550 on Build GRCh38
rs776376989 Chr.7: 139261623-139261623 on Build GRCh38
rs185500029 Chr.7: 139261608-139261608 on Build GRCh38
rs750084051 Chr.7: 139261431-139261431 on Build GRCh38
rs755391095 Chr.7: 139261557-139261557 on Build GRCh38
rs140343844 Chr.7: 139261617-139261617 on Build GRCh38
rs771169828 Chr.7: 139261473-139261473 on Build GRCh38
rs201612627 Chr.7: 139261606-139261606 on Build GRCh38
rs933109570 Chr.7: 139261535-139261535 on Build GRCh38
rs771094127 Chr.7: 139261580-139261580 on Build GRCh38
rs749924033 Chr.7: 139261514-139261514 on Build GRCh38
rs747208119 Chr.7: 139261467-139261467 on Build GRCh38
rs775212580 Chr.7: 139261474-139261474 on Build GRCh38
rs772309020 Chr.7: 139261455-139261455 on Build GRCh38
rs200979638 Chr.7: 139261598-139261598 on Build GRCh38
rs369577313 Chr.7: 139261591-139261591 on Build GRCh38
rs536452373 Chr.7: 139261573-139261573 on Build GRCh38
rs759043955 Chr.7: 139261624-139261624 on Build GRCh38
rs757051401 Chr.7: 139261542-139261542 on Build GRCh38
rs755479661 Chr.7: 139261448-139261448 on Build GRCh38
rs748616998 Chr.7: 139261451-139261451 on Build GRCh38
rs779380046 Chr.7: 139261558-139261558 on Build GRCh38
rs768625741 Chr.7: 139261482-139261482 on Build GRCh38
rs774308502 Chr.7: 139261502-139261502 on Build GRCh38
rs761675750 Chr.7: 139261506-139261506 on Build GRCh38
rs182314709 Chr.7: 139261456-139261456 on Build GRCh38
rs900177083 Chr.7: 139261487-139261487 on Build GRCh38
rs557012489 Chr.7: 139261609-139261609 on Build GRCh38
rs781168868 Chr.7: 139261557-139261558 on Build GRCh38
rs914787080 Chr.7: 139261459-139261459 on Build GRCh38
rs150369714 Chr.7: 139261618-139261618 on Build GRCh38
rs372402062 Chr.7: 139261555-139261555 on Build GRCh38
rs774077552 Chr.7: 139261607-139261607 on Build GRCh38
rs371716160 Chr.7: 139261450-139261450 on Build GRCh38
rs1002920983 Chr.7: 139261602-139261602 on Build GRCh38
rs772964991 Chr.7: 139261516-139261516 on Build GRCh38
rs754227373 Chr.7: 139261444-139261444 on Build GRCh38
rs377072849 Chr.7: 139261570-139261570 on Build GRCh38
rs147821512 Chr.7: 139261560-139261560 on Build GRCh38
rs760313627 Chr.7: 139261526-139261526 on Build GRCh38
rs368430654 Chr.7: 139261475-139261475 on Build GRCh38
rs755736823 Chr.7: 139261437-139261437 on Build GRCh38
rs958782643 Chr.7: 139261616-139261616 on Build GRCh38
rs767030609 Chr.7: 139261513-139261513 on Build GRCh38
rs780342979 Chr.7: 139261442-139261442 on Build GRCh38
rs771487652 Chr.7: 139261571-139261571 on Build GRCh38

View Full Product Details