Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 47414135-47414374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCAP
Entrez Gene ID
22937
Gene Name
SREBF chaperone
Gene Aliases
-
UniGene
Hs.531789
Gene Chromosome Location
Chr.3: 47413685-47477126 on Build GRCh38
Transcripts
NM_001320044.1 NM_012235.3 XM_005264967.1 XM_005264968.1 XM_005264971.1 XM_011533501.1 XM_011533502.2 XM_017005918.1 XM_017005919.1 XM_017005920.1 XM_017005921.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1642286 c.3571A>G p.I1191V Chr.3: 47414203-47414203 on Build GRCh38
4489204 c.3446C>T p.T1149I Chr.3: 47414328-47414328 on Build GRCh38
4489509 c.3489C>T p.V1163V Chr.3: 47414285-47414285 on Build GRCh38
7332574 c.3529G>A p.G1177S Chr.3: 47414245-47414245 on Build GRCh38
5511894 c.3449G>A p.G1150D Chr.3: 47414325-47414325 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372930525 Chr.3: 47414183-47414183 on Build GRCh38
rs1002881726 Chr.3: 47414195-47414195 on Build GRCh38
rs748207050 Chr.3: 47414319-47414319 on Build GRCh38
rs145304115 Chr.3: 47414335-47414335 on Build GRCh38
rs772461126 Chr.3: 47414174-47414174 on Build GRCh38
rs749194176 Chr.3: 47414282-47414282 on Build GRCh38
rs770327035 Chr.3: 47414320-47414320 on Build GRCh38
rs750837437 Chr.3: 47414227-47414227 on Build GRCh38
rs769512736 Chr.3: 47414164-47414165 on Build GRCh38
rs148754419 Chr.3: 47414263-47414263 on Build GRCh38
rs770868179 Chr.3: 47414293-47414293 on Build GRCh38
rs773740826 Chr.3: 47414328-47414328 on Build GRCh38
rs766784193 Chr.3: 47414351-47414351 on Build GRCh38
rs765716914 Chr.3: 47414217-47414217 on Build GRCh38
rs993859488 Chr.3: 47414175-47414175 on Build GRCh38
rs763273706 Chr.3: 47414233-47414233 on Build GRCh38
rs747259009 Chr.3: 47414296-47414296 on Build GRCh38
rs151014440 Chr.3: 47414198-47414198 on Build GRCh38
rs774515832 Chr.3: 47414354-47414354 on Build GRCh38
rs760756238 Chr.3: 47414197-47414197 on Build GRCh38
rs201231966 Chr.3: 47414265-47414265 on Build GRCh38
rs752150215 Chr.3: 47414330-47414330 on Build GRCh38
rs151202256 Chr.3: 47414317-47414317 on Build GRCh38
rs754532647 Chr.3: 47414248-47414248 on Build GRCh38
rs35628908 Chr.3: 47414240-47414240 on Build GRCh38
rs777819308 Chr.3: 47414266-47414266 on Build GRCh38
rs776507479 Chr.3: 47414201-47414201 on Build GRCh38
rs956166651 Chr.3: 47414214-47414214 on Build GRCh38
rs563706156 Chr.3: 47414241-47414241 on Build GRCh38
rs967954059 Chr.3: 47414274-47414274 on Build GRCh38
rs780762321 Chr.3: 47414254-47414254 on Build GRCh38
rs762373132 Chr.3: 47414207-47414207 on Build GRCh38
rs773147729 Chr.3: 47414191-47414193 on Build GRCh38
rs967564480 Chr.3: 47414324-47414324 on Build GRCh38
rs553479595 Chr.3: 47414173-47414173 on Build GRCh38
rs975769798 Chr.3: 47414346-47414346 on Build GRCh38
rs747973512 Chr.3: 47414163-47414164 on Build GRCh38
rs778510155 Chr.3: 47414295-47414295 on Build GRCh38

View Full Product Details