Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 73718722-73718995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB6A
Entrez Gene ID
5870
Gene Name
RAB6A, member RAS oncogene family
Gene Aliases
RAB6
UniGene
Hs.503222
Gene Chromosome Location
Chr.11: 73675638-73761156 on Build GRCh38
Transcripts
NM_001243718.1 NM_001243719.1 NM_002869.4 NM_198896.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6205065 c.221G>A p.R74H Chr.11: 73718853-73718853 on Build GRCh38
3810400 c.281A>C p.D94A Chr.11: 73718793-73718793 on Build GRCh38
294740 c.250C>T p.R84C Chr.11: 73718824-73718824 on Build GRCh38
5792224 c.209C>T p.A70V Chr.11: 73718865-73718865 on Build GRCh38
6761602 c.279C>T p.Y93Y Chr.11: 73718795-73718795 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986365604 Chr.11: 73718902-73718902 on Build GRCh38
rs111534370 Chr.11: 73718869-73718869 on Build GRCh38
rs747258009 Chr.11: 73718917-73718917 on Build GRCh38
rs907436628 Chr.11: 73718756-73718756 on Build GRCh38
rs373378980 Chr.11: 73718804-73718804 on Build GRCh38
rs776579299 Chr.11: 73718925-73718925 on Build GRCh38
rs751179397 Chr.11: 73718856-73718856 on Build GRCh38
rs780031702 Chr.11: 73718901-73718901 on Build GRCh38
rs868590451 Chr.11: 73718825-73718825 on Build GRCh38
rs752002949 Chr.11: 73718913-73718913 on Build GRCh38
rs765587579 Chr.11: 73718909-73718909 on Build GRCh38
rs375895646 Chr.11: 73718757-73718757 on Build GRCh38
rs183259590 Chr.11: 73718876-73718876 on Build GRCh38
rs878999508 Chr.11: 73718913-73718913 on Build GRCh38
rs780586189 Chr.11: 73718867-73718867 on Build GRCh38
rs773549125 Chr.11: 73718789-73718789 on Build GRCh38
rs924617406 Chr.11: 73718953-73718953 on Build GRCh38
rs770391435 Chr.11: 73718854-73718854 on Build GRCh38
rs765709332 Chr.11: 73718826-73718826 on Build GRCh38
rs768543416 Chr.11: 73718914-73718914 on Build GRCh38
rs775301035 Chr.11: 73718865-73718866 on Build GRCh38
rs370031633 Chr.11: 73718913-73718914 on Build GRCh38
rs758748500 Chr.11: 73718847-73718847 on Build GRCh38
rs763334899 Chr.11: 73718946-73718946 on Build GRCh38
rs756860407 Chr.11: 73718864-73718864 on Build GRCh38
rs371286417 Chr.11: 73718769-73718769 on Build GRCh38
rs960910171 Chr.11: 73718824-73718824 on Build GRCh38
rs528168720 Chr.11: 73718938-73718938 on Build GRCh38
rs544843396 Chr.11: 73718966-73718966 on Build GRCh38
rs747023840 Chr.11: 73718902-73718903 on Build GRCh38
rs772483425 Chr.11: 73718778-73718778 on Build GRCh38
rs559424489 Chr.11: 73718924-73718924 on Build GRCh38
rs34792668 Chr.11: 73718914-73718915 on Build GRCh38
rs773562559 Chr.11: 73718909-73718910 on Build GRCh38
rs759974645 Chr.11: 73718823-73718823 on Build GRCh38
rs749089706 Chr.11: 73718909-73718909 on Build GRCh38
rs749108970 Chr.11: 73718774-73718774 on Build GRCh38
rs760073359 Chr.11: 73718914-73718914 on Build GRCh38
rs774250337 Chr.11: 73718915-73718915 on Build GRCh38
rs1050804104 Chr.11: 73718962-73718962 on Build GRCh38
rs776042265 Chr.11: 73718932-73718932 on Build GRCh38
rs751868720 Chr.11: 73718942-73718942 on Build GRCh38
rs776901527 Chr.11: 73718903-73718903 on Build GRCh38
rs762077988 Chr.11: 73718905-73718913 on Build GRCh38
rs773416837 Chr.11: 73718927-73718927 on Build GRCh38
rs543724163 Chr.11: 73718795-73718795 on Build GRCh38
rs769718784 Chr.11: 73718767-73718767 on Build GRCh38
rs756735904 Chr.11: 73718752-73718752 on Build GRCh38
rs61758773 Chr.11: 73718840-73718840 on Build GRCh38
rs779772994 Chr.11: 73718880-73718880 on Build GRCh38
rs766827151 Chr.11: 73718911-73718913 on Build GRCh38
rs756077165 Chr.11: 73718899-73718899 on Build GRCh38
rs376485325 Chr.11: 73718809-73718809 on Build GRCh38
rs745820832 Chr.11: 73718761-73718761 on Build GRCh38

View Full Product Details