Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 167687544-167687769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDCD10
Entrez Gene ID
11235
Gene Name
programmed cell death 10
Gene Aliases
CCM3, TFAR15
UniGene
Hs.478150
Gene Chromosome Location
Chr.3: 167683891-167735690 on Build GRCh38
Transcripts
NM_007217.3 NM_145859.1 NM_145860.1 XM_005247086.4 XM_005247087.4 XM_005247088.3 XM_006713485.3 XM_011512368.2 XM_011512369.2 XM_017005644.1 XM_017005645.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6899241 c.433G>A p.V145M Chr.3: 167687656-167687656 on Build GRCh38
5665997 c.396G>T p.K132N Chr.3: 167687693-167687693 on Build GRCh38
40445 c.396G>A p.K132K Chr.3: 167687693-167687693 on Build GRCh38
1219904 c.418G>T p.E140* Chr.3: 167687671-167687671 on Build GRCh38
4333452 c.396-1G>T p.? Chr.3: 167687694-167687694 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765993541 Chr.3: 167687716-167687716 on Build GRCh38
rs886041888 Chr.3: 167687621-167687622 on Build GRCh38
rs189238762 Chr.3: 167687741-167687741 on Build GRCh38
rs757890205 Chr.3: 167687715-167687715 on Build GRCh38
rs773292567 Chr.3: 167687603-167687603 on Build GRCh38
rs185195114 Chr.3: 167687728-167687728 on Build GRCh38
rs771126809 Chr.3: 167687619-167687619 on Build GRCh38
rs751245377 Chr.3: 167687723-167687723 on Build GRCh38
rs754584968 Chr.3: 167687727-167687727 on Build GRCh38
rs760436661 Chr.3: 167687648-167687648 on Build GRCh38
rs748685407 Chr.3: 167687581-167687581 on Build GRCh38
rs367568906 Chr.3: 167687606-167687606 on Build GRCh38
rs770216793 Chr.3: 167687583-167687583 on Build GRCh38
rs367943169 Chr.3: 167687630-167687630 on Build GRCh38
rs748452548 Chr.3: 167687736-167687736 on Build GRCh38
rs776531423 Chr.3: 167687655-167687655 on Build GRCh38
rs534305257 Chr.3: 167687717-167687755 on Build GRCh38
rs749965663 Chr.3: 167687698-167687698 on Build GRCh38
rs761849146 Chr.3: 167687673-167687673 on Build GRCh38
rs976585767 Chr.3: 167687633-167687633 on Build GRCh38
rs201857174 Chr.3: 167687650-167687650 on Build GRCh38
rs765249687 Chr.3: 167687693-167687693 on Build GRCh38

View Full Product Details