Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 58492749-58492988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTMR4
Entrez Gene ID
9110
Gene Name
myotubularin related protein 4
Gene Aliases
FYVE-DSP2, ZFYVE11
UniGene
Hs.514373
Gene Chromosome Location
Chr.17: 58489532-58518109 on Build GRCh38
Transcripts
NM_004687.4 XM_005257784.2 XM_005257785.4 XM_005257786.4 XM_006722168.3 XM_011525460.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
162613 c.3219G>C p.L1073F Chr.17: 58492944-58492944 on Build GRCh38
6446211 c.3227C>T p.S1076L Chr.17: 58492936-58492936 on Build GRCh38
3820072 c.3304G>T p.D1102Y Chr.17: 58492859-58492859 on Build GRCh38
5441628 c.3229G>C p.D1077H Chr.17: 58492934-58492934 on Build GRCh38
6270270 c.3255C>A p.G1085G Chr.17: 58492908-58492908 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568743233 Chr.17: 58492801-58492801 on Build GRCh38
rs768763638 Chr.17: 58492919-58492919 on Build GRCh38
rs764050126 Chr.17: 58492950-58492950 on Build GRCh38
rs147062895 Chr.17: 58492808-58492808 on Build GRCh38
rs911603163 Chr.17: 58492782-58492782 on Build GRCh38
rs900683696 Chr.17: 58492938-58492938 on Build GRCh38
rs932880083 Chr.17: 58492816-58492816 on Build GRCh38
rs151075970 Chr.17: 58492939-58492939 on Build GRCh38
rs772094524 Chr.17: 58492897-58492897 on Build GRCh38
rs965526497 Chr.17: 58492804-58492804 on Build GRCh38
rs374266017 Chr.17: 58492828-58492828 on Build GRCh38
rs746740898 Chr.17: 58492916-58492916 on Build GRCh38
rs772506974 Chr.17: 58492790-58492790 on Build GRCh38
rs983154696 Chr.17: 58492777-58492777 on Build GRCh38
rs751876646 Chr.17: 58492792-58492792 on Build GRCh38
rs921537573 Chr.17: 58492809-58492809 on Build GRCh38
rs769957994 Chr.17: 58492953-58492953 on Build GRCh38
rs745836286 Chr.17: 58492893-58492893 on Build GRCh38
rs950524284 Chr.17: 58492775-58492775 on Build GRCh38
rs1055164069 Chr.17: 58492899-58492899 on Build GRCh38
rs377240751 Chr.17: 58492900-58492900 on Build GRCh38
rs915221371 Chr.17: 58492920-58492920 on Build GRCh38
rs778260650 Chr.17: 58492847-58492847 on Build GRCh38
rs748886288 Chr.17: 58492825-58492825 on Build GRCh38
rs761920065 Chr.17: 58492951-58492951 on Build GRCh38
rs1037390586 Chr.17: 58492927-58492927 on Build GRCh38

View Full Product Details