Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102718804-102719051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MFSD9
Entrez Gene ID
84804
Gene Name
major facilitator superfamily domain containing 9
Gene Aliases
-
UniGene
Hs.590898
Gene Chromosome Location
Chr.2: 102713867-102736888 on Build GRCh38
Transcripts
NM_001322080.1 NM_001322081.1 NM_032718.4 NR_136173.1 NR_136174.1 XM_011512005.2 XM_011512006.2 XM_011512007.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
328822 c.982A>T p.S328C Chr.2: 102718863-102718863 on Build GRCh38
5881196 c.998C>T p.A333V Chr.2: 102718847-102718847 on Build GRCh38
4561629 c.892G>A p.A298T Chr.2: 102718953-102718953 on Build GRCh38
5859799 c.1018G>A p.G340R Chr.2: 102718827-102718827 on Build GRCh38
5394057 c.958G>T p.V320L Chr.2: 102718887-102718887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144200432 Chr.2: 102719022-102719022 on Build GRCh38
rs578146378 Chr.2: 102718846-102718846 on Build GRCh38
rs769698291 Chr.2: 102718952-102718952 on Build GRCh38
rs777608154 Chr.2: 102718901-102718901 on Build GRCh38
rs757752408 Chr.2: 102719014-102719014 on Build GRCh38
rs33993717 Chr.2: 102718982-102718982 on Build GRCh38
rs372931459 Chr.2: 102718935-102718935 on Build GRCh38
rs375353978 Chr.2: 102718881-102718881 on Build GRCh38
rs201028134 Chr.2: 102719003-102719003 on Build GRCh38
rs1030240654 Chr.2: 102718993-102718993 on Build GRCh38
rs1004636483 Chr.2: 102719026-102719026 on Build GRCh38
rs748534014 Chr.2: 102718833-102718833 on Build GRCh38
rs931727775 Chr.2: 102718904-102718904 on Build GRCh38
rs573339858 Chr.2: 102719016-102719016 on Build GRCh38
rs745928094 Chr.2: 102718944-102718944 on Build GRCh38
rs751392542 Chr.2: 102718891-102718891 on Build GRCh38
rs767201966 Chr.2: 102718896-102718896 on Build GRCh38
rs781699117 Chr.2: 102719015-102719015 on Build GRCh38
rs554358456 Chr.2: 102718899-102718899 on Build GRCh38
rs148301225 Chr.2: 102718839-102718839 on Build GRCh38
rs774382444 Chr.2: 102718938-102718938 on Build GRCh38
rs904734526 Chr.2: 102718976-102718976 on Build GRCh38
rs757190136 Chr.2: 102718908-102718908 on Build GRCh38
rs753076075 Chr.2: 102718920-102718920 on Build GRCh38
rs777947369 Chr.2: 102718930-102718930 on Build GRCh38
rs750022340 Chr.2: 102718986-102718986 on Build GRCh38
rs759729239 Chr.2: 102718851-102718851 on Build GRCh38
rs139620122 Chr.2: 102718907-102718907 on Build GRCh38
rs751967614 Chr.2: 102719005-102719005 on Build GRCh38
rs781627379 Chr.2: 102718943-102718943 on Build GRCh38
rs756381972 Chr.2: 102719017-102719017 on Build GRCh38
rs781305181 Chr.2: 102718946-102718946 on Build GRCh38
rs762953843 Chr.2: 102718872-102718872 on Build GRCh38
rs779356874 Chr.2: 102718831-102718831 on Build GRCh38
rs894484528 Chr.2: 102718996-102718996 on Build GRCh38
rs375902859 Chr.2: 102718848-102718848 on Build GRCh38
rs772265230 Chr.2: 102718838-102718838 on Build GRCh38
rs762751390 Chr.2: 102718971-102718971 on Build GRCh38
rs368904822 Chr.2: 102718957-102718957 on Build GRCh38
rs200003492 Chr.2: 102718956-102718956 on Build GRCh38
rs775345876 Chr.2: 102718859-102718859 on Build GRCh38
rs377585353 Chr.2: 102718929-102718929 on Build GRCh38
rs142050183 Chr.2: 102718995-102718995 on Build GRCh38
rs771246309 Chr.2: 102718940-102718940 on Build GRCh38
rs779977008 Chr.2: 102719021-102719021 on Build GRCh38
rs34240435 Chr.2: 102718929-102718930 on Build GRCh38
rs758805278 Chr.2: 102719002-102719002 on Build GRCh38
rs986560530 Chr.2: 102719007-102719007 on Build GRCh38
rs767146863 Chr.2: 102718983-102718983 on Build GRCh38
rs768687835 Chr.2: 102718972-102718972 on Build GRCh38
rs140585828 Chr.2: 102718845-102718845 on Build GRCh38
rs749934386 Chr.2: 102718900-102718900 on Build GRCh38
rs760322503 Chr.2: 102718987-102718987 on Build GRCh38
rs765763211 Chr.2: 102718991-102718991 on Build GRCh38
rs774106067 Chr.2: 102718981-102718981 on Build GRCh38
rs769848948 Chr.2: 102718857-102718857 on Build GRCh38
rs142823158 Chr.2: 102718895-102718895 on Build GRCh38
rs768477577 Chr.2: 102719028-102719028 on Build GRCh38

View Full Product Details