Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 24140756-24141011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMC9
Entrez Gene ID
51016
Gene Name
ER membrane protein complex subunit 9
Gene Aliases
C14orf122, CGI-112, FAM158A
UniGene
Hs.271614
Gene Chromosome Location
Chr.14: 24138963-24142446 on Build GRCh38
Transcripts
NM_016049.3 XM_005267721.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5667039 c.207G>T p.V69V Chr.14: 24140957-24140957 on Build GRCh38
3814597 c.271C>G p.Q91E Chr.14: 24140893-24140893 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750527372 Chr.14: 24140867-24140867 on Build GRCh38
rs775963847 Chr.14: 24140843-24140843 on Build GRCh38
rs752418089 Chr.14: 24140881-24140881 on Build GRCh38
rs763900106 Chr.14: 24140970-24140970 on Build GRCh38
rs745409680 Chr.14: 24140902-24140902 on Build GRCh38
rs776855161 Chr.14: 24140853-24140853 on Build GRCh38
rs767486023 Chr.14: 24140869-24140869 on Build GRCh38
rs549818422 Chr.14: 24140883-24140883 on Build GRCh38
rs1011830148 Chr.14: 24140920-24140920 on Build GRCh38
rs750546145 Chr.14: 24140966-24140969 on Build GRCh38
rs146171434 Chr.14: 24140962-24140962 on Build GRCh38
rs753624343 Chr.14: 24140971-24140971 on Build GRCh38
rs766457755 Chr.14: 24140961-24140961 on Build GRCh38
rs572647668 Chr.14: 24140795-24140795 on Build GRCh38
rs761946372 Chr.14: 24140856-24140856 on Build GRCh38
rs998028248 Chr.14: 24140937-24140937 on Build GRCh38
rs765264596 Chr.14: 24140858-24140858 on Build GRCh38
rs778772783 Chr.14: 24140980-24140980 on Build GRCh38
rs765612055 Chr.14: 24140952-24140952 on Build GRCh38
rs1053340483 Chr.14: 24140892-24140892 on Build GRCh38
rs375208008 Chr.14: 24140910-24140910 on Build GRCh38
rs1054022029 Chr.14: 24140839-24140839 on Build GRCh38
rs370346086 Chr.14: 24140848-24140848 on Build GRCh38
rs369720853 Chr.14: 24140950-24140950 on Build GRCh38
rs528999680 Chr.14: 24140935-24140935 on Build GRCh38
rs563903832 Chr.14: 24140813-24140813 on Build GRCh38
rs978915531 Chr.14: 24140793-24140804 on Build GRCh38
rs755876998 Chr.14: 24140882-24140882 on Build GRCh38
rs376031934 Chr.14: 24140846-24140846 on Build GRCh38
rs541743193 Chr.14: 24140804-24140804 on Build GRCh38
rs747486156 Chr.14: 24140914-24140914 on Build GRCh38
rs764210311 Chr.14: 24140851-24140851 on Build GRCh38
rs773247890 Chr.14: 24140942-24140942 on Build GRCh38
rs778543728 Chr.14: 24140889-24140889 on Build GRCh38
rs200462788 Chr.14: 24140956-24140956 on Build GRCh38
rs964862335 Chr.14: 24140790-24140790 on Build GRCh38
rs199756160 Chr.14: 24140976-24140976 on Build GRCh38
rs892824521 Chr.14: 24140845-24140845 on Build GRCh38
rs926075195 Chr.14: 24140821-24140821 on Build GRCh38
rs754889854 Chr.14: 24140868-24140868 on Build GRCh38
rs762395253 Chr.14: 24140855-24140856 on Build GRCh38
rs561949686 Chr.14: 24140787-24140787 on Build GRCh38
rs201756898 Chr.14: 24140965-24140965 on Build GRCh38
rs373755206 Chr.14: 24140933-24140933 on Build GRCh38
rs772700123 Chr.14: 24140913-24140913 on Build GRCh38
rs745360359 Chr.14: 24140989-24140989 on Build GRCh38
rs779827416 Chr.14: 24140800-24140800 on Build GRCh38
rs200229118 Chr.14: 24140886-24140886 on Build GRCh38
rs541979784 Chr.14: 24140786-24140786 on Build GRCh38
rs141643355 Chr.14: 24140924-24140924 on Build GRCh38
rs893433614 Chr.14: 24140788-24140788 on Build GRCh38
rs374224792 Chr.14: 24140842-24140842 on Build GRCh38
rs532913523 Chr.14: 24140814-24140814 on Build GRCh38
rs768998570 Chr.14: 24140917-24140917 on Build GRCh38
rs748926411 Chr.14: 24140884-24140884 on Build GRCh38
rs934818148 Chr.14: 24140822-24140822 on Build GRCh38

View Full Product Details