Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 6009233-6009470 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIMP2
Entrez Gene ID
7965
Gene Name
aminoacyl tRNA synthetase complex interacting multifunctional protein 2
Gene Aliases
JTV-1, JTV1, P38
UniGene
Hs.301613
Gene Chromosome Location
Chr.7: 6009241-6023834 on Build GRCh38
Transcripts
NM_001326606.1 NM_001326607.1 NM_001326609.1 NM_006303.3 XM_005249848.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6488121 c.28C>G p.H10D Chr.7: 6009391-6009391 on Build GRCh38
4587498 c.49C>T p.R17C Chr.7: 6009412-6009412 on Build GRCh38
6245825 c.40G>T p.A14S Chr.7: 6009403-6009403 on Build GRCh38
4587499 c.69C>T p.C23C Chr.7: 6009432-6009432 on Build GRCh38
1091107 c.22C>A p.P8T Chr.7: 6009385-6009385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752395765 Chr.7: 6009387-6009387 on Build GRCh38
rs777401869 Chr.7: 6009393-6009393 on Build GRCh38
rs377537932 Chr.7: 6009385-6009385 on Build GRCh38
rs535443001 Chr.7: 6009268-6009268 on Build GRCh38
rs1049013110 Chr.7: 6009395-6009395 on Build GRCh38
rs182288821 Chr.7: 6009272-6009272 on Build GRCh38
rs763789143 Chr.7: 6009435-6009435 on Build GRCh38
rs756064767 Chr.7: 6009444-6009444 on Build GRCh38
rs147658637 Chr.7: 6009437-6009437 on Build GRCh38
rs566011847 Chr.7: 6009281-6009281 on Build GRCh38
rs1015568131 Chr.7: 6009252-6009252 on Build GRCh38
rs563884789 Chr.7: 6009428-6009428 on Build GRCh38
rs746850104 Chr.7: 6009396-6009396 on Build GRCh38
rs1042045384 Chr.7: 6009302-6009302 on Build GRCh38
rs142458143 Chr.7: 6009441-6009441 on Build GRCh38
rs754373311 Chr.7: 6009378-6009378 on Build GRCh38
rs911530935 Chr.7: 6009304-6009304 on Build GRCh38
rs772489394 Chr.7: 6009415-6009415 on Build GRCh38
rs780037791 Chr.7: 6009450-6009450 on Build GRCh38
rs371044703 Chr.7: 6009388-6009388 on Build GRCh38
rs970813251 Chr.7: 6009257-6009257 on Build GRCh38
rs771930297 Chr.7: 6009320-6009320 on Build GRCh38
rs746957944 Chr.7: 6009324-6009324 on Build GRCh38
rs768008472 Chr.7: 6009354-6009354 on Build GRCh38
rs145891242 Chr.7: 6009406-6009406 on Build GRCh38
rs748333627 Chr.7: 6009328-6009329 on Build GRCh38
rs762176789 Chr.7: 6009446-6009446 on Build GRCh38
rs757077360 Chr.7: 6009439-6009439 on Build GRCh38
rs766798963 Chr.7: 6009424-6009424 on Build GRCh38
rs201425558 Chr.7: 6009432-6009432 on Build GRCh38
rs140169208 Chr.7: 6009420-6009420 on Build GRCh38
rs959791854 Chr.7: 6009285-6009285 on Build GRCh38
rs570640019 Chr.7: 6009289-6009289 on Build GRCh38
rs539940178 Chr.7: 6009282-6009282 on Build GRCh38
rs767031199 Chr.7: 6009373-6009373 on Build GRCh38
rs139842556 Chr.7: 6009379-6009379 on Build GRCh38
rs769730875 Chr.7: 6009301-6009301 on Build GRCh38
rs768571855 Chr.7: 6009335-6009335 on Build GRCh38
rs761170066 Chr.7: 6009421-6009421 on Build GRCh38
rs763272580 Chr.7: 6009279-6009279 on Build GRCh38
rs202205589 Chr.7: 6009315-6009315 on Build GRCh38
rs770925960 Chr.7: 6009325-6009325 on Build GRCh38
rs748245772 Chr.7: 6009414-6009414 on Build GRCh38
rs1062372 Chr.7: 6009342-6009342 on Build GRCh38
rs945609495 Chr.7: 6009295-6009295 on Build GRCh38
rs556454943 Chr.7: 6009331-6009331 on Build GRCh38
rs149823057 Chr.7: 6009405-6009405 on Build GRCh38
rs373211005 Chr.7: 6009359-6009359 on Build GRCh38
rs558319832 Chr.7: 6009286-6009286 on Build GRCh38
rs764813325 Chr.7: 6009427-6009427 on Build GRCh38
rs777224660 Chr.7: 6009425-6009425 on Build GRCh38
rs747704354 Chr.7: 6009317-6009317 on Build GRCh38
rs768787708 Chr.7: 6009409-6009409 on Build GRCh38
rs774238548 Chr.7: 6009413-6009413 on Build GRCh38
rs745755330 Chr.7: 6009407-6009407 on Build GRCh38
rs777474354 Chr.7: 6009323-6009323 on Build GRCh38
rs772254141 Chr.7: 6009454-6009454 on Build GRCh38
rs377490622 Chr.7: 6009316-6009316 on Build GRCh38
rs915475094 Chr.7: 6009293-6009293 on Build GRCh38
rs748215903 Chr.7: 6009451-6009451 on Build GRCh38
rs574676415 Chr.7: 6009338-6009338 on Build GRCh38
rs145382336 Chr.7: 6009426-6009426 on Build GRCh38

View Full Product Details