Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 98516624-98516893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDND1
Entrez Gene ID
56650
Gene Name
claudin domain containing 1
Gene Aliases
C3orf4, GENX-3745
UniGene
Hs.531371
Gene Chromosome Location
Chr.3: 98515473-98523066 on Build GRCh38
Transcripts
NM_001040181.1 NM_001040182.1 NM_001040183.1 NM_001040199.1 NM_001040200.1 NM_019895.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4120796 c.721C>T p.R241W Chr.3: 98516700-98516700 on Build GRCh38
3735240 c.631G>A p.V211I Chr.3: 98516859-98516859 on Build GRCh38
3721267 c.756T>G p.A252A Chr.3: 98516734-98516734 on Build GRCh38
79652 c.594C>G p.L198L Chr.3: 98516827-98516827 on Build GRCh38
732212 c.560C>T p.S187L Chr.3: 98516861-98516861 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893903422 Chr.3: 98516782-98516782 on Build GRCh38
rs368555427 Chr.3: 98516811-98516811 on Build GRCh38
rs779474164 Chr.3: 98516868-98516868 on Build GRCh38
rs780932391 Chr.3: 98516858-98516858 on Build GRCh38
rs763338732 Chr.3: 98516689-98516689 on Build GRCh38
rs916374071 Chr.3: 98516688-98516688 on Build GRCh38
rs370112101 Chr.3: 98516670-98516670 on Build GRCh38
rs1004193830 Chr.3: 98516849-98516849 on Build GRCh38
rs770677430 Chr.3: 98516762-98516762 on Build GRCh38
rs878855906 Chr.3: 98516688-98516693 on Build GRCh38
rs146448772 Chr.3: 98516786-98516786 on Build GRCh38
rs372513914 Chr.3: 98516797-98516797 on Build GRCh38
rs758958263 Chr.3: 98516673-98516673 on Build GRCh38
rs755663467 Chr.3: 98516861-98516861 on Build GRCh38
rs769150314 Chr.3: 98516783-98516783 on Build GRCh38
rs745462170 Chr.3: 98516860-98516860 on Build GRCh38
rs1054266512 Chr.3: 98516803-98516803 on Build GRCh38
rs894317699 Chr.3: 98516807-98516807 on Build GRCh38
rs148992077 Chr.3: 98516793-98516793 on Build GRCh38
rs761957957 Chr.3: 98516700-98516700 on Build GRCh38
rs374891953 Chr.3: 98516683-98516683 on Build GRCh38
rs999514052 Chr.3: 98516753-98516753 on Build GRCh38
rs756116277 Chr.3: 98516728-98516728 on Build GRCh38
rs372572371 Chr.3: 98516869-98516869 on Build GRCh38
rs146171883 Chr.3: 98516746-98516746 on Build GRCh38
rs141077657 Chr.3: 98516794-98516794 on Build GRCh38
rs751452413 Chr.3: 98516846-98516846 on Build GRCh38
rs200208043 Chr.3: 98516736-98516736 on Build GRCh38
rs760831053 Chr.3: 98516804-98516804 on Build GRCh38
rs776351604 Chr.3: 98516761-98516761 on Build GRCh38
rs141729450 Chr.3: 98516709-98516709 on Build GRCh38
rs375451587 Chr.3: 98516809-98516809 on Build GRCh38
rs773236328 Chr.3: 98516771-98516771 on Build GRCh38
rs746793770 Chr.3: 98516759-98516759 on Build GRCh38
rs771953222 Chr.3: 98516663-98516663 on Build GRCh38
rs776269352 Chr.3: 98516671-98516671 on Build GRCh38
rs35785548 Chr.3: 98516782-98516783 on Build GRCh38
rs763208450 Chr.3: 98516795-98516795 on Build GRCh38
rs766035742 Chr.3: 98516654-98516655 on Build GRCh38
rs11553649 Chr.3: 98516737-98516737 on Build GRCh38
rs758277570 Chr.3: 98516743-98516743 on Build GRCh38
rs143073746 Chr.3: 98516660-98516660 on Build GRCh38
rs756974477 Chr.3: 98516850-98516850 on Build GRCh38
rs147410947 Chr.3: 98516669-98516669 on Build GRCh38
rs764394607 Chr.3: 98516699-98516699 on Build GRCh38
rs1024351133 Chr.3: 98516832-98516832 on Build GRCh38
rs746686826 Chr.3: 98516730-98516730 on Build GRCh38
rs767820286 Chr.3: 98516799-98516799 on Build GRCh38
rs767618647 Chr.3: 98516707-98516707 on Build GRCh38
rs139006614 Chr.3: 98516824-98516824 on Build GRCh38
rs140100745 Chr.3: 98516740-98516740 on Build GRCh38
rs770385121 Chr.3: 98516674-98516674 on Build GRCh38
rs750569803 Chr.3: 98516801-98516801 on Build GRCh38
rs780688644 Chr.3: 98516770-98516770 on Build GRCh38
rs147071610 Chr.3: 98516837-98516837 on Build GRCh38

View Full Product Details