Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 130268844-130269099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPA2
Entrez Gene ID
1358
Gene Name
carboxypeptidase A2
Gene Aliases
-
UniGene
Hs.490038
Gene Chromosome Location
Chr.7: 130266863-130289798 on Build GRCh38
Transcripts
NM_001869.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6860824 c.153+5T>G p.? Chr.7: 130269015-130269015 on Build GRCh38
6471630 c.79G>A p.V27I Chr.7: 130268936-130268936 on Build GRCh38
2769969 c.139C>A p.Q47K Chr.7: 130268996-130268996 on Build GRCh38
421598 c.144A>G p.E48E Chr.7: 130269001-130269001 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs780351312 Chr.7: 130268906-130268906 on Build GRCh38
rs201939124 Chr.7: 130269049-130269049 on Build GRCh38
rs776136719 Chr.7: 130268946-130268946 on Build GRCh38
rs373840499 Chr.7: 130269035-130269035 on Build GRCh38
rs1003991351 Chr.7: 130269024-130269024 on Build GRCh38
rs756637760 Chr.7: 130269006-130269006 on Build GRCh38
rs756963563 Chr.7: 130269032-130269032 on Build GRCh38
rs767535143 Chr.7: 130268879-130268879 on Build GRCh38
rs774733614 Chr.7: 130268918-130268918 on Build GRCh38
rs1033441541 Chr.7: 130269053-130269053 on Build GRCh38
rs750905965 Chr.7: 130268884-130268884 on Build GRCh38
rs200132476 Chr.7: 130268970-130268970 on Build GRCh38
rs749175052 Chr.7: 130269052-130269052 on Build GRCh38
rs777541170 Chr.7: 130268924-130268924 on Build GRCh38
rs767590420 Chr.7: 130268995-130268995 on Build GRCh38
rs755429346 Chr.7: 130269022-130269022 on Build GRCh38
rs769753562 Chr.7: 130268949-130268949 on Build GRCh38
rs754199067 Chr.7: 130269016-130269016 on Build GRCh38
rs774576113 Chr.7: 130268986-130268986 on Build GRCh38
rs756578170 Chr.7: 130268904-130268904 on Build GRCh38
rs199695765 Chr.7: 130269008-130269008 on Build GRCh38
rs746622314 Chr.7: 130268930-130268930 on Build GRCh38
rs536548762 Chr.7: 130269051-130269051 on Build GRCh38
rs778332309 Chr.7: 130269058-130269059 on Build GRCh38
rs772723641 Chr.7: 130268978-130268978 on Build GRCh38
rs750513517 Chr.7: 130269001-130269001 on Build GRCh38
rs775194258 Chr.7: 130268956-130268956 on Build GRCh38
rs779373841 Chr.7: 130269026-130269026 on Build GRCh38
rs746677107 Chr.7: 130269030-130269030 on Build GRCh38
rs769655620 Chr.7: 130269041-130269041 on Build GRCh38
rs761963942 Chr.7: 130268987-130268987 on Build GRCh38
rs763836288 Chr.7: 130268982-130268982 on Build GRCh38
rs745857648 Chr.7: 130268948-130268948 on Build GRCh38
rs900469490 Chr.7: 130268868-130268868 on Build GRCh38
rs745357813 Chr.7: 130269038-130269038 on Build GRCh38
rs762340302 Chr.7: 130268938-130268938 on Build GRCh38

View Full Product Details