Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49464064-49464309 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH16A1
Entrez Gene ID
126133
Gene Name
aldehyde dehydrogenase 16 family member A1
Gene Aliases
-
UniGene
Hs.719998
Gene Chromosome Location
Chr.19: 49453216-49471050 on Build GRCh38
Transcripts
NM_001145396.1 NM_153329.3 XM_011526441.1 XM_011526442.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6598140 c.1228C>T p.R410C Chr.19: 49464160-49464160 on Build GRCh38
2752595 c.1270C>T p.R424C Chr.19: 49464202-49464202 on Build GRCh38
3797378 c.1323G>A p.L441L Chr.19: 49464255-49464255 on Build GRCh38
1681212 c.1256C>T p.A419V Chr.19: 49464188-49464188 on Build GRCh38
7098900 c.1295G>A p.S432N Chr.19: 49464227-49464227 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs957495847 Chr.19: 49464228-49464228 on Build GRCh38
rs769307435 Chr.19: 49464203-49464203 on Build GRCh38
rs761233154 Chr.19: 49464155-49464155 on Build GRCh38
rs779005097 Chr.19: 49464183-49464183 on Build GRCh38
rs138475609 Chr.19: 49464192-49464192 on Build GRCh38
rs749386156 Chr.19: 49464118-49464118 on Build GRCh38
rs765097109 Chr.19: 49464095-49464095 on Build GRCh38
rs768925017 Chr.19: 49464275-49464275 on Build GRCh38
rs866912510 Chr.19: 49464170-49464170 on Build GRCh38
rs201780425 Chr.19: 49464111-49464111 on Build GRCh38
rs778974292 Chr.19: 49464263-49464263 on Build GRCh38
rs756773398 Chr.19: 49464094-49464094 on Build GRCh38
rs766363661 Chr.19: 49464214-49464214 on Build GRCh38
rs759331360 Chr.19: 49464234-49464234 on Build GRCh38
rs1055704325 Chr.19: 49464167-49464167 on Build GRCh38
rs369745354 Chr.19: 49464085-49464085 on Build GRCh38
rs370010620 Chr.19: 49464112-49464112 on Build GRCh38
rs143098791 Chr.19: 49464197-49464197 on Build GRCh38
rs774627148 Chr.19: 49464236-49464237 on Build GRCh38
rs1007244316 Chr.19: 49464182-49464182 on Build GRCh38
rs13346311 Chr.19: 49464240-49464240 on Build GRCh38
rs964268965 Chr.19: 49464215-49464215 on Build GRCh38
rs373500828 Chr.19: 49464266-49464266 on Build GRCh38
rs576632177 Chr.19: 49464145-49464145 on Build GRCh38
rs367630533 Chr.19: 49464280-49464280 on Build GRCh38
rs757316825 Chr.19: 49464179-49464179 on Build GRCh38
rs772769557 Chr.19: 49464191-49464191 on Build GRCh38
rs58745947 Chr.19: 49464171-49464171 on Build GRCh38
rs745923965 Chr.19: 49464189-49464189 on Build GRCh38
rs1000334154 Chr.19: 49464204-49464204 on Build GRCh38
rs150960356 Chr.19: 49464139-49464139 on Build GRCh38
rs537993303 Chr.19: 49464202-49464202 on Build GRCh38
rs757550978 Chr.19: 49464260-49464260 on Build GRCh38
rs754029074 Chr.19: 49464176-49464176 on Build GRCh38
rs746341939 Chr.19: 49464132-49464132 on Build GRCh38
rs747386854 Chr.19: 49464193-49464193 on Build GRCh38
rs780708156 Chr.19: 49464273-49464273 on Build GRCh38
rs61732836 Chr.19: 49464147-49464147 on Build GRCh38
rs753353889 Chr.19: 49464091-49464091 on Build GRCh38
rs915347915 Chr.19: 49464102-49464102 on Build GRCh38
rs1045288160 Chr.19: 49464131-49464131 on Build GRCh38
rs199807886 Chr.19: 49464227-49464227 on Build GRCh38
rs989495546 Chr.19: 49464238-49464238 on Build GRCh38
rs766273860 Chr.19: 49464161-49464161 on Build GRCh38
rs868635624 Chr.19: 49464172-49464172 on Build GRCh38
rs748810234 Chr.19: 49464284-49464284 on Build GRCh38
rs150186880 Chr.19: 49464205-49464205 on Build GRCh38
rs540098012 Chr.19: 49464285-49464285 on Build GRCh38
rs376237943 Chr.19: 49464245-49464245 on Build GRCh38
rs371448117 Chr.19: 49464281-49464281 on Build GRCh38
rs769020141 Chr.19: 49464205-49464205 on Build GRCh38
rs778054606 Chr.19: 49464117-49464117 on Build GRCh38
rs756370303 Chr.19: 49464115-49464115 on Build GRCh38
rs140640605 Chr.19: 49464144-49464144 on Build GRCh38
rs188710863 Chr.19: 49464160-49464160 on Build GRCh38
rs180723300 Chr.19: 49464274-49464274 on Build GRCh38
rs201033569 Chr.19: 49464154-49464154 on Build GRCh38
rs781655331 Chr.19: 49464278-49464278 on Build GRCh38
rs776178060 Chr.19: 49464198-49464198 on Build GRCh38
rs200435991 Chr.19: 49464164-49464164 on Build GRCh38
rs373326957 Chr.19: 49464092-49464092 on Build GRCh38
rs774728276 Chr.19: 49464124-49464124 on Build GRCh38
rs370604251 Chr.19: 49464261-49464261 on Build GRCh38
rs770986242 Chr.19: 49464123-49464123 on Build GRCh38
rs762446931 Chr.19: 49464209-49464209 on Build GRCh38
rs776285715 Chr.19: 49464152-49464152 on Build GRCh38
rs750307829 Chr.19: 49464100-49464100 on Build GRCh38
rs754754768 Chr.19: 49464163-49464163 on Build GRCh38

View Full Product Details