Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 44554484-44554731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3J
Entrez Gene ID
8669
Gene Name
eukaryotic translation initiation factor 3 subunit J
Gene Aliases
EIF3S1, eIF3-alpha, eIF3-p35
UniGene
Hs.404056
Gene Chromosome Location
Chr.15: 44537068-44562803 on Build GRCh38
Transcripts
NM_001284335.1 NM_001284336.1 NM_003758.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6227047 c.352C>G p.L118V Chr.15: 44554610-44554610 on Build GRCh38
6386030 c.319G>T p.V107L Chr.15: 44554577-44554577 on Build GRCh38
7366047 c.322C>A p.L108I Chr.15: 44554580-44554580 on Build GRCh38
6388859 c.395C>A p.A132E Chr.15: 44554653-44554653 on Build GRCh38
962105 c.355C>T p.R119W Chr.15: 44554613-44554613 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1040524878 Chr.15: 44554514-44554514 on Build GRCh38
rs369676947 Chr.15: 44554612-44554612 on Build GRCh38
rs375391650 Chr.15: 44554600-44554600 on Build GRCh38
rs34342327 Chr.15: 44554564-44554564 on Build GRCh38
rs753912691 Chr.15: 44554584-44554584 on Build GRCh38
rs748124756 Chr.15: 44554692-44554692 on Build GRCh38
rs770879138 Chr.15: 44554694-44554695 on Build GRCh38
rs772384448 Chr.15: 44554620-44554620 on Build GRCh38
rs759462070 Chr.15: 44554695-44554695 on Build GRCh38
rs759854338 Chr.15: 44554572-44554572 on Build GRCh38
rs760947528 Chr.15: 44554578-44554578 on Build GRCh38
rs767271233 Chr.15: 44554690-44554690 on Build GRCh38
rs768859327 Chr.15: 44554654-44554654 on Build GRCh38
rs376601034 Chr.15: 44554619-44554619 on Build GRCh38
rs754694847 Chr.15: 44554525-44554525 on Build GRCh38
rs767802088 Chr.15: 44554577-44554577 on Build GRCh38
rs761939111 Chr.15: 44554673-44554673 on Build GRCh38
rs1035871753 Chr.15: 44554648-44554648 on Build GRCh38
rs766383076 Chr.15: 44554587-44554587 on Build GRCh38
rs781051284 Chr.15: 44554556-44554564 on Build GRCh38
rs780663724 Chr.15: 44554529-44554529 on Build GRCh38
rs769223601 Chr.15: 44554539-44554539 on Build GRCh38
rs755725122 Chr.15: 44554708-44554708 on Build GRCh38
rs759586472 Chr.15: 44554706-44554707 on Build GRCh38
rs375912501 Chr.15: 44554513-44554513 on Build GRCh38
rs760821881 Chr.15: 44554636-44554636 on Build GRCh38
rs774515269 Chr.15: 44554705-44554706 on Build GRCh38
rs751583972 Chr.15: 44554681-44554681 on Build GRCh38
rs781005692 Chr.15: 44554597-44554597 on Build GRCh38
rs777362492 Chr.15: 44554709-44554709 on Build GRCh38
rs369516328 Chr.15: 44554674-44554674 on Build GRCh38
rs748728314 Chr.15: 44554613-44554613 on Build GRCh38
rs961564570 Chr.15: 44554653-44554653 on Build GRCh38
rs749849891 Chr.15: 44554546-44554546 on Build GRCh38
rs759509856 Chr.15: 44554685-44554685 on Build GRCh38
rs573309115 Chr.15: 44554585-44554585 on Build GRCh38
rs373067027 Chr.15: 44554614-44554614 on Build GRCh38
rs113207902 Chr.15: 44554535-44554535 on Build GRCh38
rs751426065 Chr.15: 44554590-44554590 on Build GRCh38
rs772772001 Chr.15: 44554541-44554541 on Build GRCh38
rs754775504 Chr.15: 44554596-44554596 on Build GRCh38
rs775911742 Chr.15: 44554635-44554635 on Build GRCh38
rs755775569 Chr.15: 44554610-44554610 on Build GRCh38
rs1032030578 Chr.15: 44554544-44554544 on Build GRCh38
rs764140621 Chr.15: 44554580-44554580 on Build GRCh38
rs750539473 Chr.15: 44554586-44554586 on Build GRCh38
rs746736570 Chr.15: 44554524-44554524 on Build GRCh38
rs149493333 Chr.15: 44554661-44554661 on Build GRCh38
rs34096122 Chr.15: 44554528-44554529 on Build GRCh38
rs923079644 Chr.15: 44554521-44554521 on Build GRCh38

View Full Product Details