Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 60158302-60158539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA4
Entrez Gene ID
762
Gene Name
carbonic anhydrase 4
Gene Aliases
CAIV, Car4, RP17
UniGene
Hs.89485
Gene Chromosome Location
Chr.17: 60149941-60178118 on Build GRCh38
Transcripts
XM_005257639.2 XM_011525183.2 XM_017025012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4130461 c.715C>T p.R239W Chr.17: 60158417-60158417 on Build GRCh38
6215170 c.700G>A p.V234I Chr.17: 60158402-60158402 on Build GRCh38
4994058 c.716G>A p.R239Q Chr.17: 60158418-60158418 on Build GRCh38
6319019 c.729G>T p.Q243H Chr.17: 60158431-60158431 on Build GRCh38
5594310 c.699C>T p.V233V Chr.17: 60158401-60158401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762494391 Chr.17: 60158408-60158408 on Build GRCh38
rs767868852 Chr.17: 60158389-60158389 on Build GRCh38
rs757960928 Chr.17: 60158407-60158407 on Build GRCh38
rs762909095 Chr.17: 60158375-60158375 on Build GRCh38
rs758395514 Chr.17: 60158326-60158326 on Build GRCh38
rs762619092 Chr.17: 60158515-60158515 on Build GRCh38
rs777060818 Chr.17: 60158354-60158354 on Build GRCh38
rs760920000 Chr.17: 60158486-60158486 on Build GRCh38
rs551550605 Chr.17: 60158422-60158422 on Build GRCh38
rs113977339 Chr.17: 60158447-60158447 on Build GRCh38
rs770093120 Chr.17: 60158427-60158427 on Build GRCh38
rs769858478 Chr.17: 60158358-60158358 on Build GRCh38
rs556826110 Chr.17: 60158505-60158505 on Build GRCh38
rs121434551 Chr.17: 60158357-60158357 on Build GRCh38
rs567250751 Chr.17: 60158492-60158492 on Build GRCh38
rs117704637 Chr.17: 60158402-60158402 on Build GRCh38
rs143727489 Chr.17: 60158510-60158510 on Build GRCh38
rs373253742 Chr.17: 60158449-60158449 on Build GRCh38
rs200521207 Chr.17: 60158464-60158464 on Build GRCh38
rs751538704 Chr.17: 60158415-60158415 on Build GRCh38
rs202215797 Chr.17: 60158474-60158474 on Build GRCh38
rs761553374 Chr.17: 60158398-60158398 on Build GRCh38
rs772523277 Chr.17: 60158473-60158473 on Build GRCh38
rs147852446 Chr.17: 60158399-60158399 on Build GRCh38
rs780111506 Chr.17: 60158329-60158329 on Build GRCh38
rs749440817 Chr.17: 60158463-60158463 on Build GRCh38
rs747857532 Chr.17: 60158426-60158426 on Build GRCh38
rs745324739 Chr.17: 60158321-60158321 on Build GRCh38
rs376618460 Chr.17: 60158335-60158335 on Build GRCh38
rs200131745 Chr.17: 60158470-60158470 on Build GRCh38
rs2229178 Chr.17: 60158411-60158411 on Build GRCh38
rs759956725 Chr.17: 60158388-60158388 on Build GRCh38
rs766918079 Chr.17: 60158382-60158382 on Build GRCh38
rs76995634 Chr.17: 60158418-60158418 on Build GRCh38
rs929736153 Chr.17: 60158500-60158500 on Build GRCh38
rs752951171 Chr.17: 60158390-60158390 on Build GRCh38
rs774345264 Chr.17: 60158466-60158466 on Build GRCh38
rs773370676 Chr.17: 60158364-60158364 on Build GRCh38
rs369564302 Chr.17: 60158341-60158341 on Build GRCh38
rs764759588 Chr.17: 60158401-60158401 on Build GRCh38
rs144467811 Chr.17: 60158383-60158383 on Build GRCh38

View Full Product Details