Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138284304-138284577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMC8
Entrez Gene ID
25852
Gene Name
armadillo repeat containing 8
Gene Aliases
GID5, HSPC056, S863-2, VID28
UniGene
Hs.744868
Gene Chromosome Location
Chr.3: 138187248-138298389 on Build GRCh38
Transcripts
NM_001267041.2 NM_001267042.2 NM_001282342.1 NM_015396.5 XM_006713564.3 XM_006713565.2 XM_006713566.3 XM_006713567.3 XM_011512632.1 XM_011512633.1 XM_017006095.1 XM_017006096.1 XM_017006097.1 XM_017006098.1 XM_017006099.1 NM_178130.3

Gene Symbol
NME9
Entrez Gene ID
347736
Gene Name
NME/NM23 family member 9
Gene Aliases
NM23-H9, TXL-2, TXL2, TXNDC6
UniGene
Hs.660992
Gene Chromosome Location
Chr.3: 138261437-138331785 on Build GRCh38
Transcripts
NM_001267041.2 NM_001267042.2 NM_001282342.1 NM_015396.5 XM_006713564.3 XM_006713565.2 XM_006713566.3 XM_006713567.3 XM_011512632.1 XM_011512633.1 XM_017006095.1 XM_017006096.1 XM_017006097.1 XM_017006098.1 XM_017006099.1 NM_178130.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1039010 c.1703A>G p.N568S Chr.3: 138284450-138284450 on Build GRCh38
3587949 c.1767C>T p.I589I Chr.3: 138284514-138284514 on Build GRCh38
5899424 c.1732T>A p.C578S Chr.3: 138284437-138284437 on Build GRCh38
3123885 c.1710G>A p.A570A Chr.3: 138284457-138284457 on Build GRCh38
1039011 c.1729G>T p.D577Y Chr.3: 138284476-138284476 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs956475767 Chr.3: 138284337-138284337 on Build GRCh38
rs540950609 Chr.3: 138284550-138284550 on Build GRCh38
rs961722643 Chr.3: 138284470-138284470 on Build GRCh38
rs376375211 Chr.3: 138284449-138284449 on Build GRCh38
rs370588665 Chr.3: 138284382-138284382 on Build GRCh38
rs754067445 Chr.3: 138284457-138284457 on Build GRCh38
rs371255348 Chr.3: 138284463-138284463 on Build GRCh38
rs767262292 Chr.3: 138284507-138284507 on Build GRCh38
rs755082103 Chr.3: 138284529-138284529 on Build GRCh38
rs986059701 Chr.3: 138284476-138284476 on Build GRCh38
rs75381582 Chr.3: 138284425-138284425 on Build GRCh38
rs76372822 Chr.3: 138284424-138284424 on Build GRCh38
rs77176250 Chr.3: 138284549-138284549 on Build GRCh38
rs780294149 Chr.3: 138284407-138284407 on Build GRCh38
rs78809736 Chr.3: 138284421-138284421 on Build GRCh38
rs772267910 Chr.3: 138284406-138284406 on Build GRCh38
rs374204828 Chr.3: 138284492-138284492 on Build GRCh38
rs763205943 Chr.3: 138284390-138284390 on Build GRCh38
rs775005987 Chr.3: 138284517-138284517 on Build GRCh38
rs746105710 Chr.3: 138284400-138284400 on Build GRCh38
rs763955783 Chr.3: 138284539-138284539 on Build GRCh38
rs759127728 Chr.3: 138284493-138284493 on Build GRCh38
rs374024772 Chr.3: 138284412-138284412 on Build GRCh38
rs143879950 Chr.3: 138284533-138284533 on Build GRCh38
rs914917147 Chr.3: 138284404-138284404 on Build GRCh38
rs80104568 Chr.3: 138284422-138284422 on Build GRCh38
rs544865406 Chr.3: 138284335-138284335 on Build GRCh38
rs567740683 Chr.3: 138284419-138284419 on Build GRCh38
rs911789849 Chr.3: 138284490-138284490 on Build GRCh38
rs989593758 Chr.3: 138284360-138284360 on Build GRCh38

View Full Product Details