Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140647314-140647571 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDUFA2
Entrez Gene ID
4695
Gene Name
NADH:ubiquinone oxidoreductase subunit A2
Gene Aliases
B8, CD14, CIB8
UniGene
Hs.75914
Gene Chromosome Location
Chr.5: 140645363-140647785 on Build GRCh38
Transcripts
NM_001185012.1 NM_002488.4 NR_033697.1 XM_011537663.2

Gene Symbol
TMCO6
Entrez Gene ID
55374
Gene Name
transmembrane and coiled-coil domains 6
Gene Aliases
PRO1580
UniGene
Hs.534333
Gene Chromosome Location
Chr.5: 140596529-140647411 on Build GRCh38
Transcripts
NM_001185012.1 NM_002488.4 NR_033697.1 XM_011537663.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1061730 c.123C>T p.Y41Y Chr.5: 140647341-140647341 on Build GRCh38
3850687 c.63C>T p.I21I Chr.5: 140647521-140647521 on Build GRCh38
1579450 c.74A>T p.Q25L Chr.5: 140647510-140647510 on Build GRCh38
6584554 c.23G>A p.R8Q Chr.5: 140647561-140647561 on Build GRCh38
7254630 c.42G>T p.L14L Chr.5: 140647542-140647542 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773844456 Chr.5: 140647541-140647541 on Build GRCh38
rs771180779 Chr.5: 140647428-140647428 on Build GRCh38
rs1000986821 Chr.5: 140647457-140647457 on Build GRCh38
rs751199850 Chr.5: 140647471-140647471 on Build GRCh38
rs745480793 Chr.5: 140647459-140647460 on Build GRCh38
rs553292474 Chr.5: 140647409-140647409 on Build GRCh38
rs899047509 Chr.5: 140647429-140647429 on Build GRCh38
rs773367252 Chr.5: 140647361-140647361 on Build GRCh38
rs780712775 Chr.5: 140647475-140647475 on Build GRCh38
rs757929497 Chr.5: 140647404-140647404 on Build GRCh38
rs73271536 Chr.5: 140647335-140647335 on Build GRCh38
rs745943516 Chr.5: 140647501-140647501 on Build GRCh38
rs767961780 Chr.5: 140647375-140647375 on Build GRCh38
rs753137203 Chr.5: 140647376-140647376 on Build GRCh38
rs201963018 Chr.5: 140647373-140647373 on Build GRCh38
rs1052307009 Chr.5: 140647441-140647441 on Build GRCh38
rs963295784 Chr.5: 140647492-140647492 on Build GRCh38
rs779660717 Chr.5: 140647408-140647408 on Build GRCh38
rs1022558681 Chr.5: 140647552-140647552 on Build GRCh38
rs994836172 Chr.5: 140647434-140647434 on Build GRCh38
rs769050771 Chr.5: 140647518-140647518 on Build GRCh38
rs752496272 Chr.5: 140647478-140647478 on Build GRCh38
rs879089727 Chr.5: 140647480-140647480 on Build GRCh38
rs866793910 Chr.5: 140647436-140647436 on Build GRCh38
rs878965270 Chr.5: 140647481-140647481 on Build GRCh38
rs764517073 Chr.5: 140647388-140647388 on Build GRCh38
rs886552960 Chr.5: 140647444-140647444 on Build GRCh38
rs763129842 Chr.5: 140647367-140647367 on Build GRCh38
rs777120962 Chr.5: 140647526-140647526 on Build GRCh38
rs775860662 Chr.5: 140647449-140647449 on Build GRCh38
rs1044297835 Chr.5: 140647400-140647400 on Build GRCh38
rs574408648 Chr.5: 140647421-140647421 on Build GRCh38
rs749897719 Chr.5: 140647402-140647402 on Build GRCh38
rs746760218 Chr.5: 140647340-140647340 on Build GRCh38
rs371682984 Chr.5: 140647490-140647490 on Build GRCh38
rs948525047 Chr.5: 140647391-140647391 on Build GRCh38
rs754726383 Chr.5: 140647412-140647412 on Build GRCh38
rs563223074 Chr.5: 140647504-140647504 on Build GRCh38
rs878948076 Chr.5: 140647479-140647479 on Build GRCh38
rs766668017 Chr.5: 140647371-140647371 on Build GRCh38
rs900714849 Chr.5: 140647401-140647401 on Build GRCh38
rs538560846 Chr.5: 140647489-140647489 on Build GRCh38
rs765942108 Chr.5: 140647465-140647465 on Build GRCh38
rs200503970 Chr.5: 140647456-140647456 on Build GRCh38
rs558825457 Chr.5: 140647359-140647359 on Build GRCh38
rs899800350 Chr.5: 140647453-140647453 on Build GRCh38
rs371250205 Chr.5: 140647433-140647433 on Build GRCh38
rs777882601 Chr.5: 140647426-140647426 on Build GRCh38
rs774681546 Chr.5: 140647431-140647431 on Build GRCh38
rs762420223 Chr.5: 140647464-140647464 on Build GRCh38
rs752340193 Chr.5: 140647545-140647545 on Build GRCh38
rs779189461 Chr.5: 140647497-140647497 on Build GRCh38
rs777827533 Chr.5: 140647488-140647488 on Build GRCh38
rs1014802286 Chr.5: 140647482-140647482 on Build GRCh38
rs768546550 Chr.5: 140647346-140647346 on Build GRCh38
rs772307877 Chr.5: 140647446-140647446 on Build GRCh38
rs201522225 Chr.5: 140647374-140647374 on Build GRCh38
rs771125764 Chr.5: 140647491-140647491 on Build GRCh38
rs879022054 Chr.5: 140647437-140647437 on Build GRCh38
rs748184129 Chr.5: 140647355-140647355 on Build GRCh38
rs754602793 Chr.5: 140647474-140647474 on Build GRCh38
rs755904736 Chr.5: 140647548-140647548 on Build GRCh38
rs776402455 Chr.5: 140647354-140647354 on Build GRCh38
rs755882393 Chr.5: 140647487-140647487 on Build GRCh38
rs996216750 Chr.5: 140647435-140647435 on Build GRCh38
rs35672074 Chr.5: 140647544-140647544 on Build GRCh38
rs761081473 Chr.5: 140647505-140647505 on Build GRCh38
rs765747363 Chr.5: 140647539-140647539 on Build GRCh38
rs761189881 Chr.5: 140647384-140647384 on Build GRCh38
rs781099916 Chr.5: 140647420-140647420 on Build GRCh38
rs769705612 Chr.5: 140647424-140647424 on Build GRCh38
rs754394533 Chr.5: 140647461-140647461 on Build GRCh38
rs375905956 Chr.5: 140647553-140647553 on Build GRCh38

View Full Product Details