Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 163512063-163512302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAT2B
Entrez Gene ID
27430
Gene Name
methionine adenosyltransferase 2B
Gene Aliases
MAT-II, MATIIbeta, Nbla02999, SDR23E1, TGR
UniGene
Hs.54642
Gene Chromosome Location
Chr.5: 163503064-163519353 on Build GRCh38
Transcripts
NM_013283.4 NM_182796.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3248067 c.164C>T p.A55V Chr.5: 163512102-163512102 on Build GRCh38
3248066 c.131C>T p.A44V Chr.5: 163512102-163512102 on Build GRCh38
4141620 c.126G>T p.W42C Chr.5: 163512097-163512097 on Build GRCh38
5402961 c.233T>C p.V78A Chr.5: 163512171-163512171 on Build GRCh38
312764 c.215A>G p.H72R Chr.5: 163512186-163512186 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376437565 Chr.5: 163512220-163512220 on Build GRCh38
rs140671288 Chr.5: 163512245-163512245 on Build GRCh38
rs778177105 Chr.5: 163512154-163512154 on Build GRCh38
rs201956239 Chr.5: 163512185-163512185 on Build GRCh38
rs757849549 Chr.5: 163512234-163512234 on Build GRCh38
rs151289006 Chr.5: 163512225-163512225 on Build GRCh38
rs779701901 Chr.5: 163512250-163512250 on Build GRCh38
rs375517945 Chr.5: 163512252-163512252 on Build GRCh38
rs761739816 Chr.5: 163512219-163512219 on Build GRCh38
rs756473772 Chr.5: 163512111-163512111 on Build GRCh38
rs186526604 Chr.5: 163512166-163512166 on Build GRCh38
rs77036976 Chr.5: 163512253-163512253 on Build GRCh38
rs749215967 Chr.5: 163512263-163512264 on Build GRCh38
rs886490464 Chr.5: 163512251-163512251 on Build GRCh38
rs754307442 Chr.5: 163512229-163512229 on Build GRCh38
rs370978931 Chr.5: 163512228-163512228 on Build GRCh38
rs768446603 Chr.5: 163512189-163512189 on Build GRCh38
rs369618222 Chr.5: 163512099-163512099 on Build GRCh38
rs746809727 Chr.5: 163512188-163512188 on Build GRCh38
rs1039350502 Chr.5: 163512203-163512203 on Build GRCh38
rs745377555 Chr.5: 163512172-163512172 on Build GRCh38
rs370330528 Chr.5: 163512190-163512190 on Build GRCh38
rs143459228 Chr.5: 163512169-163512169 on Build GRCh38
rs759670860 Chr.5: 163512222-163512222 on Build GRCh38
rs779380579 Chr.5: 163512244-163512244 on Build GRCh38
rs369992014 Chr.5: 163512227-163512227 on Build GRCh38
rs780022492 Chr.5: 163512212-163512212 on Build GRCh38
rs751613594 Chr.5: 163512174-163512174 on Build GRCh38
rs970615499 Chr.5: 163512264-163512264 on Build GRCh38
rs766388761 Chr.5: 163512093-163512093 on Build GRCh38
rs757863904 Chr.5: 163512248-163512248 on Build GRCh38
rs767834890 Chr.5: 163512104-163512104 on Build GRCh38

View Full Product Details