Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 61412091-61412352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDH20
Entrez Gene ID
64881
Gene Name
protocadherin 20
Gene Aliases
PCDH13
UniGene
Hs.391781
Gene Chromosome Location
Chr.13: 61408963-61427266 on Build GRCh38
Transcripts
NM_022843.3 XM_017020708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
696341 c.1921G>T p.A641S Chr.13: 61412097-61412097 on Build GRCh38
4880678 c.1721A>T p.E574V Chr.13: 61412297-61412297 on Build GRCh38
4533258 c.1879G>A p.G627S Chr.13: 61412139-61412139 on Build GRCh38
4048215 c.1726G>T p.E576* Chr.13: 61412292-61412292 on Build GRCh38
4904525 c.1846G>A p.D616N Chr.13: 61412172-61412172 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774405422 Chr.13: 61412242-61412242 on Build GRCh38
rs759236518 Chr.13: 61412155-61412155 on Build GRCh38
rs866507512 Chr.13: 61412179-61412179 on Build GRCh38
rs770989899 Chr.13: 61412133-61412133 on Build GRCh38
rs926675800 Chr.13: 61412138-61412138 on Build GRCh38
rs775371578 Chr.13: 61412159-61412159 on Build GRCh38
rs758932068 Chr.13: 61412192-61412192 on Build GRCh38
rs753042731 Chr.13: 61412303-61412303 on Build GRCh38
rs758845398 Chr.13: 61412322-61412322 on Build GRCh38
rs767094941 Chr.13: 61412283-61412283 on Build GRCh38
rs764054697 Chr.13: 61412169-61412169 on Build GRCh38
rs980817543 Chr.13: 61412132-61412132 on Build GRCh38
rs575590366 Chr.13: 61412178-61412178 on Build GRCh38
rs200308516 Chr.13: 61412221-61412221 on Build GRCh38
rs762744072 Chr.13: 61412235-61412235 on Build GRCh38
rs746999849 Chr.13: 61412212-61412212 on Build GRCh38
rs778346963 Chr.13: 61412200-61412200 on Build GRCh38
rs771098462 Chr.13: 61412215-61412215 on Build GRCh38
rs770029803 Chr.13: 61412232-61412232 on Build GRCh38
rs760354256 Chr.13: 61412293-61412293 on Build GRCh38
rs866914126 Chr.13: 61412121-61412121 on Build GRCh38
rs878906311 Chr.13: 61412310-61412310 on Build GRCh38
rs775229877 Chr.13: 61412234-61412234 on Build GRCh38
rs762832042 Chr.13: 61412163-61412163 on Build GRCh38
rs1039026882 Chr.13: 61412246-61412246 on Build GRCh38
rs545200706 Chr.13: 61412143-61412143 on Build GRCh38
rs767064828 Chr.13: 61412181-61412181 on Build GRCh38
rs779468628 Chr.13: 61412187-61412187 on Build GRCh38
rs750969895 Chr.13: 61412170-61412170 on Build GRCh38
rs201960619 Chr.13: 61412115-61412115 on Build GRCh38
rs34196441 Chr.13: 61412284-61412284 on Build GRCh38
rs781466949 Chr.13: 61412186-61412186 on Build GRCh38
rs778125916 Chr.13: 61412324-61412324 on Build GRCh38
rs752173190 Chr.13: 61412325-61412325 on Build GRCh38
rs868405292 Chr.13: 61412141-61412141 on Build GRCh38
rs769638860 Chr.13: 61412158-61412158 on Build GRCh38
rs144379684 Chr.13: 61412173-61412173 on Build GRCh38
rs149653186 Chr.13: 61412167-61412167 on Build GRCh38
rs753134724 Chr.13: 61412188-61412188 on Build GRCh38
rs769936521 Chr.13: 61412286-61412286 on Build GRCh38
rs1047883713 Chr.13: 61412125-61412125 on Build GRCh38
rs766161151 Chr.13: 61412299-61412299 on Build GRCh38

View Full Product Details