Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34752366-34752617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJD2
Entrez Gene ID
57369
Gene Name
gap junction protein delta 2
Gene Aliases
CX36, GJA9
UniGene
Hs.283816
Gene Chromosome Location
Chr.15: 34751746-34754955 on Build GRCh38
Transcripts
NM_020660.2 XM_017022438.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6481064 c.857G>C p.G286A Chr.15: 34752587-34752587 on Build GRCh38
4855395 c.893G>A p.R298H Chr.15: 34752551-34752551 on Build GRCh38
416512 c.886G>A p.E296K Chr.15: 34752558-34752558 on Build GRCh38
960870 c.853C>T p.R285* Chr.15: 34752591-34752591 on Build GRCh38
6142304 c.847G>T p.A283S Chr.15: 34752597-34752597 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770815406 Chr.15: 34752522-34752522 on Build GRCh38
rs544517317 Chr.15: 34752431-34752431 on Build GRCh38
rs753637450 Chr.15: 34752595-34752595 on Build GRCh38
rs201362920 Chr.15: 34752409-34752409 on Build GRCh38
rs558322044 Chr.15: 34752400-34752400 on Build GRCh38
rs1056318330 Chr.15: 34752538-34752538 on Build GRCh38
rs780972179 Chr.15: 34752458-34752458 on Build GRCh38
rs200667131 Chr.15: 34752429-34752429 on Build GRCh38
rs202009924 Chr.15: 34752533-34752533 on Build GRCh38
rs199863427 Chr.15: 34752424-34752424 on Build GRCh38
rs768992622 Chr.15: 34752538-34752538 on Build GRCh38
rs749201223 Chr.15: 34752514-34752514 on Build GRCh38
rs754307386 Chr.15: 34752441-34752441 on Build GRCh38
rs1038772684 Chr.15: 34752446-34752446 on Build GRCh38
rs937912303 Chr.15: 34752529-34752529 on Build GRCh38
rs199744979 Chr.15: 34752410-34752410 on Build GRCh38
rs201852436 Chr.15: 34752415-34752415 on Build GRCh38
rs759110061 Chr.15: 34752569-34752569 on Build GRCh38
rs766013958 Chr.15: 34752448-34752448 on Build GRCh38
rs751078368 Chr.15: 34752450-34752450 on Build GRCh38
rs372564429 Chr.15: 34752451-34752451 on Build GRCh38
rs775572644 Chr.15: 34752551-34752551 on Build GRCh38
rs147648302 Chr.15: 34752536-34752536 on Build GRCh38
rs757724805 Chr.15: 34752445-34752445 on Build GRCh38
rs955978248 Chr.15: 34752469-34752469 on Build GRCh38
rs577090680 Chr.15: 34752425-34752425 on Build GRCh38
rs772340286 Chr.15: 34752541-34752541 on Build GRCh38
rs908550461 Chr.15: 34752414-34752414 on Build GRCh38
rs777460358 Chr.15: 34752590-34752590 on Build GRCh38
rs767065723 Chr.15: 34752571-34752571 on Build GRCh38
rs373145560 Chr.15: 34752438-34752438 on Build GRCh38
rs200785650 Chr.15: 34752586-34752586 on Build GRCh38
rs896285359 Chr.15: 34752542-34752542 on Build GRCh38
rs201005605 Chr.15: 34752413-34752413 on Build GRCh38
rs200915359 Chr.15: 34752481-34752481 on Build GRCh38
rs138585940 Chr.15: 34752582-34752582 on Build GRCh38
rs34964522 Chr.15: 34752556-34752556 on Build GRCh38
rs774453640 Chr.15: 34752524-34752524 on Build GRCh38
rs762443981 Chr.15: 34752566-34752566 on Build GRCh38
rs752301576 Chr.15: 34752585-34752585 on Build GRCh38
rs150150700 Chr.15: 34752560-34752560 on Build GRCh38
rs755862258 Chr.15: 34752461-34752461 on Build GRCh38
rs751166485 Chr.15: 34752568-34752568 on Build GRCh38
rs765811174 Chr.15: 34752567-34752567 on Build GRCh38
rs777010107 Chr.15: 34752562-34752562 on Build GRCh38

View Full Product Details