Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141151506-141151773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHB6
Entrez Gene ID
56130
Gene Name
protocadherin beta 6
Gene Aliases
PCDH-BETA6
UniGene
Hs.283085
Gene Chromosome Location
Chr.5: 141150022-141153287 on Build GRCh38
Transcripts
NM_001303145.1 NM_018939.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1696396 c.1294C>T p.Q432* Chr.5: 141151551-141151551 on Build GRCh38
5867575 c.1358C>T p.S453F Chr.5: 141151615-141151615 on Build GRCh38
3919012 c.1347C>T p.F449F Chr.5: 141151604-141151604 on Build GRCh38
4382543 c.1321G>A p.D441N Chr.5: 141151578-141151578 on Build GRCh38
6169575 c.1460C>G p.S487W Chr.5: 141151717-141151717 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139417417 Chr.5: 141151691-141151691 on Build GRCh38
rs144860035 Chr.5: 141151629-141151629 on Build GRCh38
rs782566190 Chr.5: 141151554-141151554 on Build GRCh38
rs782324654 Chr.5: 141151586-141151586 on Build GRCh38
rs782768292 Chr.5: 141151736-141151736 on Build GRCh38
rs781900353 Chr.5: 141151635-141151635 on Build GRCh38
rs782013587 Chr.5: 141151671-141151671 on Build GRCh38
rs782069574 Chr.5: 141151628-141151628 on Build GRCh38
rs246707 Chr.5: 141151594-141151594 on Build GRCh38
rs782019131 Chr.5: 141151741-141151741 on Build GRCh38
rs782445585 Chr.5: 141151637-141151637 on Build GRCh38
rs782687648 Chr.5: 141151663-141151663 on Build GRCh38
rs199862013 Chr.5: 141151576-141151576 on Build GRCh38
rs782112984 Chr.5: 141151734-141151734 on Build GRCh38
rs374320078 Chr.5: 141151670-141151670 on Build GRCh38
rs782003526 Chr.5: 141151608-141151608 on Build GRCh38
rs149392351 Chr.5: 141151612-141151612 on Build GRCh38
rs782230409 Chr.5: 141151590-141151590 on Build GRCh38
rs147347959 Chr.5: 141151675-141151675 on Build GRCh38
rs782362726 Chr.5: 141151660-141151661 on Build GRCh38
rs782105322 Chr.5: 141151668-141151668 on Build GRCh38
rs782267489 Chr.5: 141151715-141151715 on Build GRCh38
rs150523954 Chr.5: 141151731-141151731 on Build GRCh38
rs782574464 Chr.5: 141151646-141151646 on Build GRCh38
rs781818708 Chr.5: 141151648-141151648 on Build GRCh38
rs544845940 Chr.5: 141151752-141151752 on Build GRCh38
rs782170627 Chr.5: 141151583-141151583 on Build GRCh38
rs782694959 Chr.5: 141151673-141151673 on Build GRCh38
rs201339779 Chr.5: 141151677-141151677 on Build GRCh38
rs781852986 Chr.5: 141151694-141151694 on Build GRCh38
rs782160583 Chr.5: 141151749-141151749 on Build GRCh38
rs782698019 Chr.5: 141151566-141151566 on Build GRCh38
rs61745657 Chr.5: 141151696-141151696 on Build GRCh38
rs781906903 Chr.5: 141151698-141151698 on Build GRCh38
rs377527452 Chr.5: 141151543-141151543 on Build GRCh38
rs139574362 Chr.5: 141151733-141151733 on Build GRCh38
rs370052306 Chr.5: 141151623-141151623 on Build GRCh38
rs782623021 Chr.5: 141151573-141151573 on Build GRCh38
rs781816634 Chr.5: 141151570-141151570 on Build GRCh38
rs782219760 Chr.5: 141151653-141151654 on Build GRCh38
rs781924526 Chr.5: 141151624-141151624 on Build GRCh38
rs202237118 Chr.5: 141151580-141151580 on Build GRCh38
rs782440656 Chr.5: 141151699-141151699 on Build GRCh38
rs782649380 Chr.5: 141151711-141151711 on Build GRCh38
rs782062278 Chr.5: 141151727-141151727 on Build GRCh38
rs782588902 Chr.5: 141151700-141151700 on Build GRCh38
rs782426060 Chr.5: 141151718-141151718 on Build GRCh38
rs146356849 Chr.5: 141151577-141151577 on Build GRCh38
rs782185954 Chr.5: 141151664-141151664 on Build GRCh38
rs246706 Chr.5: 141151634-141151634 on Build GRCh38
rs781915601 Chr.5: 141151724-141151724 on Build GRCh38
rs782477871 Chr.5: 141151572-141151572 on Build GRCh38
rs782683075 Chr.5: 141151582-141151582 on Build GRCh38
rs377458637 Chr.5: 141151703-141151703 on Build GRCh38
rs781946790 Chr.5: 141151588-141151588 on Build GRCh38
rs782158956 Chr.5: 141151672-141151672 on Build GRCh38
rs782265707 Chr.5: 141151660-141151660 on Build GRCh38
rs782361782 Chr.5: 141151701-141151701 on Build GRCh38
rs532016504 Chr.5: 141151533-141151533 on Build GRCh38
rs782478636 Chr.5: 141151650-141151650 on Build GRCh38
rs17844436 Chr.5: 141151680-141151680 on Build GRCh38
rs199646293 Chr.5: 141151633-141151633 on Build GRCh38
rs782330858 Chr.5: 141151665-141151665 on Build GRCh38
rs200017004 Chr.5: 141151654-141151654 on Build GRCh38
rs782527550 Chr.5: 141151695-141151695 on Build GRCh38
rs769508807 Chr.5: 141151667-141151667 on Build GRCh38
rs782174509 Chr.5: 141151709-141151709 on Build GRCh38
rs573025846 Chr.5: 141151662-141151662 on Build GRCh38
rs34727235 Chr.5: 141151688-141151688 on Build GRCh38
rs782639686 Chr.5: 141151659-141151659 on Build GRCh38

View Full Product Details