Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23061724-23061997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RSPH14
Entrez Gene ID
27156
Gene Name
radial spoke head 14 homolog
Gene Aliases
RTDR1
UniGene
Hs.526920
Gene Chromosome Location
Chr.22: 23059408-23180048 on Build GRCh38
Transcripts
NM_014433.2 XM_011530149.2 XM_011530150.1 XM_011530151.2 XM_011530152.2 XM_011530154.2 XM_011530155.2 XM_017028774.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6770180 c.757G>A p.G253S Chr.22: 23061842-23061842 on Build GRCh38
1307995 c.696C>T p.V232V Chr.22: 23061903-23061903 on Build GRCh38
6770179 c.750C>T p.N250N Chr.22: 23061849-23061849 on Build GRCh38
6770178 c.690delT p.F230fs*11 Chr.22: 23061909-23061909 on Build GRCh38
5586448 c.706C>T p.L236L Chr.22: 23061893-23061893 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150054974 Chr.22: 23061840-23061840 on Build GRCh38
rs989389326 Chr.22: 23061959-23061959 on Build GRCh38
rs199729382 Chr.22: 23061842-23061842 on Build GRCh38
rs546811807 Chr.22: 23061868-23061868 on Build GRCh38
rs761588812 Chr.22: 23061802-23061802 on Build GRCh38
rs764746692 Chr.22: 23061918-23061918 on Build GRCh38
rs530022296 Chr.22: 23061843-23061843 on Build GRCh38
rs1016683509 Chr.22: 23061864-23061864 on Build GRCh38
rs941583483 Chr.22: 23061878-23061878 on Build GRCh38
rs753918821 Chr.22: 23061821-23061821 on Build GRCh38
rs113209847 Chr.22: 23061789-23061789 on Build GRCh38
rs574436787 Chr.22: 23061755-23061755 on Build GRCh38
rs528801226 Chr.22: 23061788-23061788 on Build GRCh38
rs775728745 Chr.22: 23061888-23061888 on Build GRCh38
rs191338653 Chr.22: 23061860-23061860 on Build GRCh38
rs558861150 Chr.22: 23061950-23061950 on Build GRCh38
rs954571763 Chr.22: 23061749-23061749 on Build GRCh38
rs371291239 Chr.22: 23061906-23061906 on Build GRCh38
rs778340707 Chr.22: 23061775-23061775 on Build GRCh38
rs766332003 Chr.22: 23061790-23061791 on Build GRCh38
rs200796554 Chr.22: 23061956-23061956 on Build GRCh38
rs781735205 Chr.22: 23061846-23061846 on Build GRCh38
rs756623949 Chr.22: 23061908-23061908 on Build GRCh38
rs775160616 Chr.22: 23061797-23061797 on Build GRCh38
rs768003828 Chr.22: 23061905-23061905 on Build GRCh38
rs377321870 Chr.22: 23061869-23061869 on Build GRCh38
rs774209305 Chr.22: 23061801-23061801 on Build GRCh38
rs111977700 Chr.22: 23061957-23061957 on Build GRCh38
rs747996426 Chr.22: 23061863-23061863 on Build GRCh38
rs748858884 Chr.22: 23061922-23061922 on Build GRCh38
rs907275410 Chr.22: 23061910-23061910 on Build GRCh38
rs769355704 Chr.22: 23061856-23061856 on Build GRCh38
rs200251269 Chr.22: 23061796-23061796 on Build GRCh38
rs757668979 Chr.22: 23061844-23061844 on Build GRCh38
rs180969312 Chr.22: 23061933-23061933 on Build GRCh38
rs776416957 Chr.22: 23061793-23061793 on Build GRCh38
rs935252205 Chr.22: 23061780-23061780 on Build GRCh38
rs775673673 Chr.22: 23061970-23061970 on Build GRCh38
rs543235904 Chr.22: 23061757-23061757 on Build GRCh38
rs780504357 Chr.22: 23061792-23061792 on Build GRCh38
rs765472813 Chr.22: 23061885-23061885 on Build GRCh38
rs748881554 Chr.22: 23061859-23061859 on Build GRCh38
rs893155840 Chr.22: 23061973-23061973 on Build GRCh38
rs747865701 Chr.22: 23061948-23061948 on Build GRCh38
rs779470437 Chr.22: 23061914-23061914 on Build GRCh38
rs186403758 Chr.22: 23061798-23061798 on Build GRCh38
rs567774889 Chr.22: 23061849-23061849 on Build GRCh38
rs566627157 Chr.22: 23061899-23061899 on Build GRCh38
rs959517109 Chr.22: 23061794-23061794 on Build GRCh38
rs767519941 Chr.22: 23061808-23061808 on Build GRCh38
rs759739722 Chr.22: 23061822-23061822 on Build GRCh38
rs954788442 Chr.22: 23061938-23061938 on Build GRCh38
rs770999138 Chr.22: 23061960-23061960 on Build GRCh38
rs117662831 Chr.22: 23061769-23061769 on Build GRCh38
rs757677516 Chr.22: 23061904-23061904 on Build GRCh38
rs764412189 Chr.22: 23061895-23061895 on Build GRCh38
rs763021987 Chr.22: 23061893-23061893 on Build GRCh38
rs778675971 Chr.22: 23061944-23061944 on Build GRCh38
rs759148115 Chr.22: 23061964-23061965 on Build GRCh38
rs530778842 Chr.22: 23061828-23061828 on Build GRCh38
rs760483020 Chr.22: 23061787-23061789 on Build GRCh38
rs199783550 Chr.22: 23061832-23061832 on Build GRCh38

View Full Product Details