Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12702053-12702278 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNPO2
Entrez Gene ID
30000
Gene Name
transportin 2
Gene Aliases
IPO3, KPNB2B, TRN2
UniGene
Hs.416049
Gene Chromosome Location
Chr.19: 12699194-12723996 on Build GRCh38
Transcripts
NM_001136195.1 NM_001136196.1 NM_013433.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7289013 c.2398T>C p.F800L Chr.19: 12702085-12702085 on Build GRCh38
165043 c.2374C>T p.R792W Chr.19: 12702079-12702079 on Build GRCh38
7289014 c.2368T>C p.F790L Chr.19: 12702085-12702085 on Build GRCh38
991411 c.2314A>G p.T772A Chr.19: 12702169-12702169 on Build GRCh38
4809915 c.2404C>T p.R802W Chr.19: 12702079-12702079 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372019164 Chr.19: 12702134-12702134 on Build GRCh38
rs769255420 Chr.19: 12702178-12702178 on Build GRCh38
rs780055965 Chr.19: 12702159-12702159 on Build GRCh38
rs563324567 Chr.19: 12702208-12702208 on Build GRCh38
rs376478097 Chr.19: 12702137-12702137 on Build GRCh38
rs920346744 Chr.19: 12702242-12702242 on Build GRCh38
rs775989389 Chr.19: 12702198-12702198 on Build GRCh38
rs764218305 Chr.19: 12702121-12702121 on Build GRCh38
rs767909855 Chr.19: 12702110-12702110 on Build GRCh38
rs754669083 Chr.19: 12702156-12702156 on Build GRCh38
rs754683714 Chr.19: 12702216-12702216 on Build GRCh38
rs757223735 Chr.19: 12702122-12702122 on Build GRCh38
rs756416352 Chr.19: 12702115-12702115 on Build GRCh38
rs765443392 Chr.19: 12702210-12702210 on Build GRCh38
rs759927883 Chr.19: 12702089-12702089 on Build GRCh38
rs370026661 Chr.19: 12702164-12702164 on Build GRCh38
rs374413929 Chr.19: 12702167-12702167 on Build GRCh38
rs767997823 Chr.19: 12702197-12702197 on Build GRCh38
rs368882556 Chr.19: 12702174-12702174 on Build GRCh38
rs745756507 Chr.19: 12702136-12702136 on Build GRCh38
rs201401387 Chr.19: 12702083-12702083 on Build GRCh38
rs1027641431 Chr.19: 12702098-12702098 on Build GRCh38
rs994790985 Chr.19: 12702097-12702097 on Build GRCh38
rs747717622 Chr.19: 12702175-12702175 on Build GRCh38
rs367890401 Chr.19: 12702086-12702086 on Build GRCh38
rs760085512 Chr.19: 12702192-12702192 on Build GRCh38
rs760879225 Chr.19: 12702202-12702202 on Build GRCh38
rs116518190 Chr.19: 12702168-12702168 on Build GRCh38
rs757386951 Chr.19: 12702207-12702207 on Build GRCh38
rs545804517 Chr.19: 12702213-12702213 on Build GRCh38
rs753038373 Chr.19: 12702113-12702113 on Build GRCh38
rs912472017 Chr.19: 12702220-12702220 on Build GRCh38
rs746837120 Chr.19: 12702214-12702214 on Build GRCh38
rs764389543 Chr.19: 12702203-12702203 on Build GRCh38
rs549891121 Chr.19: 12702212-12702212 on Build GRCh38
rs372789391 Chr.19: 12702093-12702093 on Build GRCh38
rs772678471 Chr.19: 12702187-12702187 on Build GRCh38

View Full Product Details