Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 130159949-130160198 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST14
Entrez Gene ID
6768
Gene Name
suppression of tumorigenicity 14
Gene Aliases
ARCI11, HAI, MT-SP1, MTSP1, PRSS14, SNC19, TADG15, TMPRSS14
UniGene
Hs.504315
Gene Chromosome Location
Chr.11: 130159787-130210362 on Build GRCh38
Transcripts
NM_021978.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6045713 c.31G>A p.G11R Chr.11: 130160010-130160010 on Build GRCh38
7276319 c.64T>C p.Y22H Chr.11: 130160043-130160043 on Build GRCh38
7282162 c.35_36insC p.K14fs*86 Chr.11: 130160014-130160015 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1056912126 Chr.11: 130160021-130160021 on Build GRCh38
rs370051145 Chr.11: 130159985-130159985 on Build GRCh38
rs372702262 Chr.11: 130160000-130160000 on Build GRCh38
rs555935688 Chr.11: 130160116-130160116 on Build GRCh38
rs923589815 Chr.11: 130160132-130160132 on Build GRCh38
rs1001016375 Chr.11: 130160135-130160135 on Build GRCh38
rs942833502 Chr.11: 130160156-130160156 on Build GRCh38
rs748673581 Chr.11: 130160038-130160038 on Build GRCh38
rs762651539 Chr.11: 130160077-130160077 on Build GRCh38
rs960518835 Chr.11: 130160083-130160083 on Build GRCh38
rs770708446 Chr.11: 130160105-130160105 on Build GRCh38
rs377743342 Chr.11: 130160155-130160155 on Build GRCh38
rs979091550 Chr.11: 130160130-130160130 on Build GRCh38
rs756155146 Chr.11: 130160074-130160074 on Build GRCh38
rs1039952164 Chr.11: 130160176-130160176 on Build GRCh38
rs1050794374 Chr.11: 130160149-130160149 on Build GRCh38
rs567582561 Chr.11: 130159991-130159991 on Build GRCh38
rs868109235 Chr.11: 130160055-130160055 on Build GRCh38
rs542010199 Chr.11: 130160012-130160012 on Build GRCh38
rs1039899816 Chr.11: 130160011-130160011 on Build GRCh38
rs773765762 Chr.11: 130160075-130160075 on Build GRCh38
rs765303362 Chr.11: 130160069-130160069 on Build GRCh38
rs904093978 Chr.11: 130160126-130160126 on Build GRCh38
rs1026164636 Chr.11: 130160023-130160023 on Build GRCh38
rs968135902 Chr.11: 130160119-130160119 on Build GRCh38
rs758642514 Chr.11: 130159977-130159977 on Build GRCh38
rs137852932 Chr.11: 130159982-130159982 on Build GRCh38
rs1029799664 Chr.11: 130160157-130160157 on Build GRCh38
rs781717480 Chr.11: 130160015-130160015 on Build GRCh38
rs951527318 Chr.11: 130160034-130160034 on Build GRCh38
rs865979295 Chr.11: 130159999-130159999 on Build GRCh38
rs3740883 Chr.11: 130160143-130160143 on Build GRCh38
rs897278826 Chr.11: 130159974-130159974 on Build GRCh38
rs955826592 Chr.11: 130160148-130160148 on Build GRCh38

View Full Product Details