Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 124891258-124891507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ROBO4
Entrez Gene ID
54538
Gene Name
roundabout guidance receptor 4
Gene Aliases
ECSM4, MRB
UniGene
Hs.524121
Gene Chromosome Location
Chr.11: 124884218-124897935 on Build GRCh38
Transcripts
NM_001301088.1 NM_019055.5 XM_006718861.1 XM_011542875.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
924451 c.1749C>T p.G583G Chr.11: 124891498-124891498 on Build GRCh38
6768343 c.1817G>A p.R606H Chr.11: 124891430-124891430 on Build GRCh38
5435258 c.1781C>A p.P594Q Chr.11: 124891466-124891466 on Build GRCh38
1659067 c.1926T>C p.A642A Chr.11: 124891321-124891321 on Build GRCh38
458700 c.1758C>G p.I586M Chr.11: 124891489-124891489 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374820251 Chr.11: 124891296-124891296 on Build GRCh38
rs146643213 Chr.11: 124891332-124891332 on Build GRCh38
rs767884863 Chr.11: 124891288-124891288 on Build GRCh38
rs751371459 Chr.11: 124891422-124891422 on Build GRCh38
rs1041512303 Chr.11: 124891449-124891449 on Build GRCh38
rs145535350 Chr.11: 124891347-124891347 on Build GRCh38
rs376195962 Chr.11: 124891398-124891398 on Build GRCh38
rs772195431 Chr.11: 124891458-124891458 on Build GRCh38
rs143995311 Chr.11: 124891472-124891472 on Build GRCh38
rs749338809 Chr.11: 124891431-124891431 on Build GRCh38
rs763628320 Chr.11: 124891297-124891297 on Build GRCh38
rs746033291 Chr.11: 124891448-124891448 on Build GRCh38
rs779759158 Chr.11: 124891407-124891407 on Build GRCh38
rs758624847 Chr.11: 124891391-124891393 on Build GRCh38
rs570388613 Chr.11: 124891299-124891299 on Build GRCh38
rs762249155 Chr.11: 124891482-124891482 on Build GRCh38
rs750049739 Chr.11: 124891385-124891385 on Build GRCh38
rs774678606 Chr.11: 124891367-124891367 on Build GRCh38
rs958298127 Chr.11: 124891324-124891324 on Build GRCh38
rs750650495 Chr.11: 124891386-124891387 on Build GRCh38
rs764775703 Chr.11: 124891400-124891400 on Build GRCh38
rs764754815 Chr.11: 124891388-124891388 on Build GRCh38
rs778886357 Chr.11: 124891444-124891444 on Build GRCh38
rs946298494 Chr.11: 124891476-124891476 on Build GRCh38
rs767880271 Chr.11: 124891371-124891371 on Build GRCh38
rs920470915 Chr.11: 124891369-124891369 on Build GRCh38
rs753204104 Chr.11: 124891291-124891291 on Build GRCh38
rs751878563 Chr.11: 124891486-124891486 on Build GRCh38
rs750044288 Chr.11: 124891401-124891401 on Build GRCh38
rs369176966 Chr.11: 124891310-124891310 on Build GRCh38
rs766671399 Chr.11: 124891440-124891446 on Build GRCh38
rs777326650 Chr.11: 124891475-124891475 on Build GRCh38
rs982252695 Chr.11: 124891353-124891353 on Build GRCh38
rs746911218 Chr.11: 124891334-124891334 on Build GRCh38
rs556036732 Chr.11: 124891302-124891302 on Build GRCh38
rs770990130 Chr.11: 124891442-124891442 on Build GRCh38
rs977349745 Chr.11: 124891445-124891445 on Build GRCh38
rs200121967 Chr.11: 124891460-124891460 on Build GRCh38
rs372455722 Chr.11: 124891361-124891361 on Build GRCh38
rs149507368 Chr.11: 124891430-124891430 on Build GRCh38
rs537736034 Chr.11: 124891301-124891301 on Build GRCh38
rs138111911 Chr.11: 124891383-124891383 on Build GRCh38
rs759696358 Chr.11: 124891368-124891368 on Build GRCh38
rs758018710 Chr.11: 124891402-124891402 on Build GRCh38
rs761186797 Chr.11: 124891468-124891468 on Build GRCh38
rs140544911 Chr.11: 124891346-124891346 on Build GRCh38
rs944578139 Chr.11: 124891434-124891434 on Build GRCh38

View Full Product Details