Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 159780826-159781097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCP1
Entrez Gene ID
6950
Gene Name
t-complex 1
Gene Aliases
CCT-alpha, CCT1, CCTa, D6S230E, TCP-1-alpha
UniGene
Hs.363137
Gene Chromosome Location
Chr.6: 159778498-159789703 on Build GRCh38
Transcripts
NM_001008897.1 NM_030752.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3023358 c.914C>A p.A305D Chr.6: 159780994-159780994 on Build GRCh38
3941613 c.859C>G p.L287V Chr.6: 159781049-159781049 on Build GRCh38
5869697 c.913G>T p.A305S Chr.6: 159780995-159780995 on Build GRCh38
7211273 c.900T>C p.F300F Chr.6: 159781008-159781008 on Build GRCh38
5420604 c.973+1G>T p.? Chr.6: 159780934-159780934 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780520451 Chr.6: 159780928-159780928 on Build GRCh38
rs750832526 Chr.6: 159780916-159780916 on Build GRCh38
rs759379389 Chr.6: 159780886-159780886 on Build GRCh38
rs762711626 Chr.6: 159781014-159781014 on Build GRCh38
rs750964687 Chr.6: 159781047-159781047 on Build GRCh38
rs369732107 Chr.6: 159780976-159780977 on Build GRCh38
rs146349098 Chr.6: 159781040-159781040 on Build GRCh38
rs952787922 Chr.6: 159780991-159780991 on Build GRCh38
rs752431453 Chr.6: 159780942-159780942 on Build GRCh38
rs769607726 Chr.6: 159780990-159780990 on Build GRCh38
rs768920093 Chr.6: 159781017-159781017 on Build GRCh38
rs756516873 Chr.6: 159780917-159780917 on Build GRCh38
rs768140073 Chr.6: 159780914-159780914 on Build GRCh38
rs2295900 Chr.6: 159780863-159780863 on Build GRCh38
rs767442680 Chr.6: 159781046-159781046 on Build GRCh38
rs766963421 Chr.6: 159781049-159781049 on Build GRCh38
rs917431116 Chr.6: 159781056-159781056 on Build GRCh38
rs565289489 Chr.6: 159780889-159780889 on Build GRCh38
rs745741436 Chr.6: 159780976-159780976 on Build GRCh38
rs775710793 Chr.6: 159780893-159780893 on Build GRCh38
rs561544024 Chr.6: 159781055-159781055 on Build GRCh38
rs984087056 Chr.6: 159780999-159780999 on Build GRCh38
rs908794210 Chr.6: 159781033-159781033 on Build GRCh38
rs756814026 Chr.6: 159781069-159781069 on Build GRCh38
rs142939243 Chr.6: 159780956-159780956 on Build GRCh38
rs992941356 Chr.6: 159781052-159781052 on Build GRCh38
rs748157457 Chr.6: 159781044-159781044 on Build GRCh38
rs775301824 Chr.6: 159781012-159781012 on Build GRCh38
rs759967605 Chr.6: 159781026-159781028 on Build GRCh38
rs777545486 Chr.6: 159780955-159780955 on Build GRCh38
rs777637046 Chr.6: 159781057-159781057 on Build GRCh38
rs763119502 Chr.6: 159780895-159780895 on Build GRCh38
rs921371219 Chr.6: 159780972-159780972 on Build GRCh38
rs373745424 Chr.6: 159780911-159780911 on Build GRCh38
rs1007686076 Chr.6: 159780861-159780861 on Build GRCh38
rs781202599 Chr.6: 159780967-159780967 on Build GRCh38
rs780966412 Chr.6: 159780887-159780887 on Build GRCh38
rs1009401299 Chr.6: 159780882-159780882 on Build GRCh38
rs764267916 Chr.6: 159780899-159780899 on Build GRCh38
rs758167068 Chr.6: 159780946-159780946 on Build GRCh38
rs770280174 Chr.6: 159780965-159780965 on Build GRCh38
rs180680763 Chr.6: 159780871-159780871 on Build GRCh38
rs776420429 Chr.6: 159780885-159780885 on Build GRCh38
rs754305861 Chr.6: 159781051-159781051 on Build GRCh38

View Full Product Details