Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7620372-7620633 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XAB2
Entrez Gene ID
56949
Gene Name
XPA binding protein 2
Gene Aliases
HCNP, HCRN, NTC90, SYF1
UniGene
Hs.9822
Gene Chromosome Location
Chr.19: 7619525-7629553 on Build GRCh38
Transcripts
NM_020196.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6823670 c.2117C>T p.T706M Chr.19: 7620424-7620424 on Build GRCh38
4535186 c.2164G>A p.E722K Chr.19: 7620377-7620377 on Build GRCh38
394697 c.2106G>T p.A702A Chr.19: 7620435-7620435 on Build GRCh38
1711705 c.2057C>T p.A686V Chr.19: 7620584-7620584 on Build GRCh38
4741569 c.2092delC p.R698fs*72 Chr.19: 7620549-7620549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776207900 Chr.19: 7620582-7620582 on Build GRCh38
rs772151574 Chr.19: 7620474-7620474 on Build GRCh38
rs749406741 Chr.19: 7620527-7620527 on Build GRCh38
rs994199794 Chr.19: 7620588-7620588 on Build GRCh38
rs937438657 Chr.19: 7620515-7620516 on Build GRCh38
rs375084353 Chr.19: 7620463-7620463 on Build GRCh38
rs763507107 Chr.19: 7620578-7620578 on Build GRCh38
rs750266829 Chr.19: 7620514-7620514 on Build GRCh38
rs763933385 Chr.19: 7620584-7620585 on Build GRCh38
rs779224839 Chr.19: 7620591-7620591 on Build GRCh38
rs755765150 Chr.19: 7620407-7620407 on Build GRCh38
rs199646554 Chr.19: 7620444-7620444 on Build GRCh38
rs771200676 Chr.19: 7620536-7620536 on Build GRCh38
rs149923081 Chr.19: 7620568-7620568 on Build GRCh38
rs768841987 Chr.19: 7620438-7620438 on Build GRCh38
rs758371805 Chr.19: 7620517-7620517 on Build GRCh38
rs776915758 Chr.19: 7620441-7620441 on Build GRCh38
rs4134865 Chr.19: 7620437-7620437 on Build GRCh38
rs373345053 Chr.19: 7620464-7620464 on Build GRCh38
rs746992256 Chr.19: 7620521-7620521 on Build GRCh38
rs775949931 Chr.19: 7620574-7620574 on Build GRCh38
rs377349471 Chr.19: 7620450-7620450 on Build GRCh38
rs780109493 Chr.19: 7620520-7620520 on Build GRCh38
rs200696126 Chr.19: 7620496-7620496 on Build GRCh38
rs201877455 Chr.19: 7620594-7620594 on Build GRCh38
rs778503828 Chr.19: 7620511-7620511 on Build GRCh38
rs544589268 Chr.19: 7620470-7620470 on Build GRCh38
rs78349437 Chr.19: 7620501-7620501 on Build GRCh38
rs756980443 Chr.19: 7620471-7620471 on Build GRCh38
rs760105715 Chr.19: 7620455-7620455 on Build GRCh38
rs758245097 Chr.19: 7620607-7620607 on Build GRCh38
rs770034942 Chr.19: 7620478-7620478 on Build GRCh38
rs752368471 Chr.19: 7620455-7620456 on Build GRCh38
rs374854321 Chr.19: 7620505-7620505 on Build GRCh38
rs753963390 Chr.19: 7620412-7620412 on Build GRCh38
rs375490958 Chr.19: 7620495-7620495 on Build GRCh38
rs201887776 Chr.19: 7620465-7620465 on Build GRCh38
rs781374520 Chr.19: 7620476-7620476 on Build GRCh38
rs763708971 Chr.19: 7620502-7620502 on Build GRCh38
rs554681666 Chr.19: 7620472-7620472 on Build GRCh38
rs748269855 Chr.19: 7620477-7620477 on Build GRCh38
rs746259500 Chr.19: 7620549-7620549 on Build GRCh38
rs770164799 Chr.19: 7620445-7620445 on Build GRCh38
rs200379177 Chr.19: 7620500-7620500 on Build GRCh38
rs1038500272 Chr.19: 7620533-7620533 on Build GRCh38
rs750141778 Chr.19: 7620604-7620604 on Build GRCh38
rs371192037 Chr.19: 7620423-7620423 on Build GRCh38
rs372386074 Chr.19: 7620485-7620485 on Build GRCh38
rs747797751 Chr.19: 7620404-7620404 on Build GRCh38
rs780740847 Chr.19: 7620403-7620403 on Build GRCh38
rs370847959 Chr.19: 7620468-7620468 on Build GRCh38
rs1006150405 Chr.19: 7620397-7620397 on Build GRCh38
rs758318567 Chr.19: 7620472-7620473 on Build GRCh38
rs769754079 Chr.19: 7620524-7620524 on Build GRCh38
rs753419342 Chr.19: 7620504-7620504 on Build GRCh38
rs774785093 Chr.19: 7620490-7620490 on Build GRCh38
rs761228150 Chr.19: 7620577-7620577 on Build GRCh38
rs777979258 Chr.19: 7620526-7620526 on Build GRCh38
rs370348644 Chr.19: 7620453-7620453 on Build GRCh38
rs139206953 Chr.19: 7620411-7620411 on Build GRCh38
rs770857451 Chr.19: 7620435-7620435 on Build GRCh38
rs759456580 Chr.19: 7620598-7620598 on Build GRCh38
rs574809769 Chr.19: 7620479-7620479 on Build GRCh38
rs117645581 Chr.19: 7620424-7620424 on Build GRCh38
rs773587070 Chr.19: 7620446-7620446 on Build GRCh38
rs970854911 Chr.19: 7620592-7620592 on Build GRCh38
rs771198353 Chr.19: 7620486-7620486 on Build GRCh38
rs774527896 Chr.19: 7620539-7620539 on Build GRCh38
rs368794166 Chr.19: 7620475-7620475 on Build GRCh38
rs766948905 Chr.19: 7620458-7620458 on Build GRCh38

View Full Product Details