Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 72975500-72975769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFYVE1
Entrez Gene ID
53349
Gene Name
zinc finger FYVE-type containing 1
Gene Aliases
DFCP1, PPP1R172, SR3, TAFF1, ZNFN2A1
UniGene
Hs.335106
Gene Chromosome Location
Chr.14: 72969445-73027212 on Build GRCh38
Transcripts
NM_001281734.1 NM_001281735.1 NM_021260.3 NM_178441.2 XM_017021373.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7422614 c.1663A>T p.N555Y Chr.14: 72975694-72975694 on Build GRCh38
117976 c.1729G>T p.G577* Chr.14: 72975628-72975628 on Build GRCh38
6463645 c.1687G>A p.G563R Chr.14: 72975670-72975670 on Build GRCh38
3377451 c.1658A>G p.N553S Chr.14: 72975699-72975699 on Build GRCh38
214182 c.1637C>T p.T546I Chr.14: 72975720-72975720 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147680028 Chr.14: 72975699-72975699 on Build GRCh38
rs574741894 Chr.14: 72975741-72975741 on Build GRCh38
rs145340873 Chr.14: 72975647-72975647 on Build GRCh38
rs767418945 Chr.14: 72975648-72975648 on Build GRCh38
rs762821060 Chr.14: 72975632-72975632 on Build GRCh38
rs774938921 Chr.14: 72975631-72975631 on Build GRCh38
rs1018821967 Chr.14: 72975709-72975709 on Build GRCh38
rs556307289 Chr.14: 72975735-72975735 on Build GRCh38
rs745324866 Chr.14: 72975583-72975583 on Build GRCh38
rs923121876 Chr.14: 72975740-72975740 on Build GRCh38
rs374928375 Chr.14: 72975641-72975641 on Build GRCh38
rs935299870 Chr.14: 72975623-72975623 on Build GRCh38
rs776334086 Chr.14: 72975543-72975543 on Build GRCh38
rs772679418 Chr.14: 72975521-72975521 on Build GRCh38
rs139201234 Chr.14: 72975671-72975671 on Build GRCh38
rs903826574 Chr.14: 72975593-72975593 on Build GRCh38
rs538270310 Chr.14: 72975720-72975720 on Build GRCh38
rs772294626 Chr.14: 72975681-72975681 on Build GRCh38
rs900092433 Chr.14: 72975728-72975728 on Build GRCh38
rs760423155 Chr.14: 72975586-72975586 on Build GRCh38
rs370445582 Chr.14: 72975592-72975592 on Build GRCh38
rs984761748 Chr.14: 72975626-72975626 on Build GRCh38
rs750537981 Chr.14: 72975670-72975670 on Build GRCh38
rs528390491 Chr.14: 72975539-72975539 on Build GRCh38
rs201983681 Chr.14: 72975640-72975640 on Build GRCh38
rs139314162 Chr.14: 72975638-72975638 on Build GRCh38
rs753945348 Chr.14: 72975734-72975734 on Build GRCh38

View Full Product Details