Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 58671093-58671362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STX16
Entrez Gene ID
8675
Gene Name
syntaxin 16
Gene Aliases
SYN16
UniGene
Hs.307913
Gene Chromosome Location
Chr.20: 58651253-58679526 on Build GRCh38
Transcripts
NM_001001433.2 NM_001134772.2 NM_001134773.2 NM_001204868.1 NM_003763.5 NR_037941.1 NR_037942.1 NR_037943.1 NR_037945.1

Gene Symbol
STX16-NPEPL1
Entrez Gene ID
100534593
Gene Name
STX16-NPEPL1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.654868
Gene Chromosome Location
Chr.20: 58651253-58715844 on Build GRCh38
Transcripts
NM_001001433.2 NM_001134772.2 NM_001134773.2 NM_001204868.1 NM_003763.5 NR_037941.1 NR_037942.1 NR_037943.1 NR_037945.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6046938 c.713G>A p.R238Q Chr.20: 58671218-58671218 on Build GRCh38
4099780 c.715G>C p.E239Q Chr.20: 58671220-58671220 on Build GRCh38
4932816 c.594A>G p.T198T Chr.20: 58671162-58671162 on Build GRCh38
4099783 c.769T>G p.L257V Chr.20: 58671274-58671274 on Build GRCh38
4915334 c.593C>A p.T198K Chr.20: 58671161-58671161 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151312981 Chr.20: 58671287-58671287 on Build GRCh38
rs748600539 Chr.20: 58671261-58671261 on Build GRCh38
rs371863588 Chr.20: 58671281-58671281 on Build GRCh38
rs546565420 Chr.20: 58671162-58671162 on Build GRCh38
rs765198026 Chr.20: 58671321-58671321 on Build GRCh38
rs771102032 Chr.20: 58671282-58671282 on Build GRCh38
rs763050618 Chr.20: 58671198-58671198 on Build GRCh38
rs751180587 Chr.20: 58671252-58671252 on Build GRCh38
rs202102691 Chr.20: 58671221-58671221 on Build GRCh38
rs748582456 Chr.20: 58671167-58671167 on Build GRCh38
rs200589976 Chr.20: 58671190-58671190 on Build GRCh38
rs772070373 Chr.20: 58671303-58671303 on Build GRCh38
rs754524705 Chr.20: 58671256-58671256 on Build GRCh38
rs764248085 Chr.20: 58671331-58671332 on Build GRCh38
rs769324515 Chr.20: 58671165-58671165 on Build GRCh38
rs776576965 Chr.20: 58671311-58671311 on Build GRCh38
rs756533057 Chr.20: 58671265-58671265 on Build GRCh38
rs758353477 Chr.20: 58671226-58671226 on Build GRCh38
rs1031420787 Chr.20: 58671286-58671286 on Build GRCh38
rs148806774 Chr.20: 58671227-58671227 on Build GRCh38
rs142393313 Chr.20: 58671228-58671228 on Build GRCh38
rs766194853 Chr.20: 58671325-58671325 on Build GRCh38
rs535191941 Chr.20: 58671268-58671268 on Build GRCh38
rs115536505 Chr.20: 58671212-58671212 on Build GRCh38
rs776141349 Chr.20: 58671141-58671141 on Build GRCh38
rs781763936 Chr.20: 58671166-58671166 on Build GRCh38
rs147330346 Chr.20: 58671218-58671218 on Build GRCh38
rs983488302 Chr.20: 58671145-58671145 on Build GRCh38
rs761607016 Chr.20: 58671318-58671318 on Build GRCh38
rs188308261 Chr.20: 58671211-58671211 on Build GRCh38
rs766023155 Chr.20: 58671234-58671234 on Build GRCh38
rs754541084 Chr.20: 58671327-58671327 on Build GRCh38
rs910176347 Chr.20: 58671136-58671136 on Build GRCh38
rs6100159 Chr.20: 58671297-58671297 on Build GRCh38
rs757957533 Chr.20: 58671125-58671125 on Build GRCh38
rs773488877 Chr.20: 58671195-58671195 on Build GRCh38
rs762802079 Chr.20: 58671324-58671324 on Build GRCh38
rs745958261 Chr.20: 58671288-58671288 on Build GRCh38
rs759584494 Chr.20: 58671217-58671217 on Build GRCh38
rs781768372 Chr.20: 58671257-58671257 on Build GRCh38
rs767096253 Chr.20: 58671331-58671331 on Build GRCh38
rs779381461 Chr.20: 58671130-58671130 on Build GRCh38
rs1014720820 Chr.20: 58671283-58671283 on Build GRCh38
rs202209533 Chr.20: 58671279-58671279 on Build GRCh38
rs763187247 Chr.20: 58671325-58671325 on Build GRCh38

View Full Product Details