Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 32324927-32325152 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRCA2
Entrez Gene ID
675
Gene Name
BRCA2, DNA repair associated
Gene Aliases
BRCC2, BROVCA2, FACD, FAD, FAD1, FANCD, FANCD1, GLM3, PNCA2, XRCC11
UniGene
Hs.34012
Gene Chromosome Location
Chr.13: 32315480-32399672 on Build GRCh38
Transcripts
NM_000059.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3786166 c.317-10A>G p.? Chr.13: 32325066-32325066 on Build GRCh38
7245259 c.346A>T p.S116C Chr.13: 32325105-32325105 on Build GRCh38
3955533 c.346A>C p.S116R Chr.13: 32325105-32325105 on Build GRCh38
469350 c.374A>G p.D125G Chr.13: 32325133-32325133 on Build GRCh38
7400281 c.355A>G p.T119A Chr.13: 32325114-32325114 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs80358586 Chr.13: 32325100-32325100 on Build GRCh38
rs80358603 Chr.13: 32325112-32325112 on Build GRCh38
rs786203060 Chr.13: 32325098-32325098 on Build GRCh38
rs370259333 Chr.13: 32325041-32325041 on Build GRCh38
rs397507680 Chr.13: 32325123-32325123 on Build GRCh38
rs397507309 Chr.13: 32325107-32325107 on Build GRCh38
rs869320790 Chr.13: 32325092-32325092 on Build GRCh38
rs369065391 Chr.13: 32325029-32325029 on Build GRCh38
rs587778129 Chr.13: 32325099-32325099 on Build GRCh38
rs56242644 Chr.13: 32325102-32325102 on Build GRCh38
rs775484431 Chr.13: 32325047-32325047 on Build GRCh38
rs548654788 Chr.13: 32324978-32324978 on Build GRCh38
rs564490783 Chr.13: 32325024-32325024 on Build GRCh38
rs766194968 Chr.13: 32325109-32325109 on Build GRCh38
rs80358567 Chr.13: 32325081-32325081 on Build GRCh38
rs778029839 Chr.13: 32325039-32325039 on Build GRCh38
rs81002841 Chr.13: 32325064-32325064 on Build GRCh38
rs81002794 Chr.13: 32325054-32325054 on Build GRCh38
rs928238003 Chr.13: 32324963-32324963 on Build GRCh38
rs781377719 Chr.13: 32325049-32325049 on Build GRCh38
rs772010158 Chr.13: 32325083-32325083 on Build GRCh38
rs773184912 Chr.13: 32325094-32325094 on Build GRCh38
rs81002891 Chr.13: 32325022-32325022 on Build GRCh38
rs375125172 Chr.13: 32325111-32325111 on Build GRCh38
rs878853568 Chr.13: 32325101-32325102 on Build GRCh38
rs111623805 Chr.13: 32325010-32325010 on Build GRCh38
rs929618744 Chr.13: 32324996-32324996 on Build GRCh38
rs876659161 Chr.13: 32325097-32325097 on Build GRCh38
rs81002824 Chr.13: 32325066-32325066 on Build GRCh38
rs969483891 Chr.13: 32325036-32325036 on Build GRCh38
rs758910027 Chr.13: 32325011-32325011 on Build GRCh38
rs11571609 Chr.13: 32324980-32324980 on Build GRCh38
rs80358568 Chr.13: 32325082-32325082 on Build GRCh38

View Full Product Details